Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

91986

Millipore

PALCAM Listeria Selective Supplement according to Van Netten et al.

suitable for microbiology

Synonym(s):

Listeria Selective Supplement (PALCAM)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41171614
NACRES:
NA.85

sterility

sterile

form

powder

shelf life

limited shelf life, expiry date on the label

application(s)

environmental
food and beverages

microbiology

storage temp.

2-8°C

suitability

Listeria spp.

General description

An antimicrobial supplement recommended for the selective isolation and identification of Listeria monocytogenes.

Application

for the selective isolation and identification of Listeria monocytogenes

Components

(per vial, sufficient for 500 ml medium)
Ceftazidime 10.0 mg
Polymyxin B 50000 IU
Acriflavin-hydrochloride 2.5 mg

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 3 - Eye Dam. 1 - Resp. Sens. 1 - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service