Skip to Content
MilliporeSigma

1.12122

PALCAM Listeria selective supplement

for Listeria spp., pkg of 10 vials, for use with PALCAM Listeria Selective Agar Base, for use with L-PALCAM Listeria Selective Enrichment Broth Base

Synonym(s):

Listeria selective supplement

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

NACRES:
NA.74
UNSPSC Code:
41106212
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

form

medium granules (dehydrated (DCM))

composition

Acriflavine, 2.5 mg per vial , Ceftazidime, 10 mg per vial , Polymixin-B-sulfate, 5 mg per vial

packaging

pkg of 10 vials

technique(s)

microbiological culture: suitable

application(s)

food and beverages
microbiology
pathogen testing

compatibility

for use with L-PALCAM Listeria Selective Enrichment Broth Base , for use with PALCAM Listeria Selective Agar Base

suitability

Listeria spp.

General description

PALCAM Listeria Selective Supplement is a mixture of three antibiotics in lyophilized form. It inhibits the growth of accompanying bacteria in the selective cultivation of Listeria monocytogenes.

Application

PALCAM Listeria Selective Supplement is an additive for the preparation of PALCAM Listeria Selective Agar (1.11755) and L-PALCAM Listeria-Selective Enrichment Broth (Base) (1.10823).

Analysis Note

Supplement tested in PALCAM-Listeria-Selektive agar (Base) acc. to VAN NETTEN et al. art. no. 1.11755.
Recovery on test medium
- Listeria monocytogenes ATCC 13932 (WDCM 00021):
≥ 50 %
- Listeria monocytogenes ATCC 19111 (WDCM 00020): ≥ 50 %
- Listeria monocytogenes ATCC 35152 (WDCM 00109): ≥ 50 %
- Listeria innocua ATCC 33090 (WDCM 00017): ≥ 50 %
- Staphylococcus aureus ATCC 6538 (WDCM 00032): ≤ 0.01 %
- Enterococcus faecalis ATCC 19433 (WDCM 00009): ≤ 0.01 %
- Escherichia coli ATCC 25922 (WDCM 00013): ≤ 0.01 %
Black zone
- Listeria monocytogenes ATCC 13932 (WDCM 00021):
+
- Listeria monocytogenes ATCC 19111 (WDCM 00020): +
- Listeria monocytogenes ATCC 35152 (WDCM 00109): +
- Listeria innocua ATCC 33090 (WDCM 00017): +
Incubation: Listeria 48 hrs.; 35 °C; aerobaic. Other strains 72 hrs.; 30 °C; aerobic.

Other Notes

Replaces: 1.12122.0017


Pictograms

Health hazardCorrosion

Signal Word

Danger

Hazard Classifications

Aquatic Chronic 3 - Eye Dam. 1 - Resp. Sens. 1 - Skin Sens. 1

Storage Class

11 - Combustible Solids

WGK

WGK 3



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library