Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

91558

Supelco

Methyl tridecanoate

analytical standard

Synonym(s):

Tridecanoic acid methyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)11COOCH3
CAS Number:
Molecular Weight:
228.37
Beilstein:
1769695
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24
Pricing and availability is not currently available.

grade

analytical standard

Quality Level

Assay

≥99.0% (GC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

refractive index

n20/D 1.435 (lit.)
n20/D 1.435

bp

131 °C/4 mmHg (lit.)

mp

5.5 °C (lit.)

density

0.864 g/mL at 25 °C (lit.)

format

neat

functional group

ester

shipped in

ambient

storage temp.

room temp

SMILES string

CCCCCCCCCCCCC(=O)OC

InChI

1S/C14H28O2/c1-3-4-5-6-7-8-9-10-11-12-13-14(15)16-2/h3-13H2,1-2H3

InChI key

JNDDPBOKWCBQSM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Methyl tridecanoate has been used as an internal standard for the gas chromatographic (GC) analysis of the total lipids obtained from Chlamydomonas reinhardtii microalgae containing a thioesterase enzyme from Dunaliella. tertiolecta.[1]
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Chronic 4 - Eye Dam. 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Expression of the heterologous Dunaliella tertiolecta fatty acyl-ACP thioesterase leads to increased lipid production in Chlamydomonas reinhardtii
Tan MWK and Lee KY
Journal of Biotechnology, 247, 60-67 (2017)
Samira Asghari et al.
Journal of cellular physiology, 234(7), 12011-12018 (2018-12-06)
Despite at the beginning known as a benign disease, endometriosis is defined as a risk factor for developing ovarian carcinoma. The effect of endometriosis on ovarian malignancy is known but its role in granulosa cell tumor is still unclear. In
Oier Aizpurua-Olaizola et al.
Journal of food science, 80(1), E101-E107 (2014-12-05)
In this study, supercritical fluid extraction has been successfully applied to a sequential fractionation of fatty acids and polyphenols from wine wastes (2 different vitis vinifera grapes). To this aim, in a 1st step just fatty acids were extracted and

Protocols

Protocol for GC Analysis of Bacterial Acid Methyl Esters (BAMEs) on Equity®-1

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service