Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

61689

Supelco

Methyl dodecanoate

analytical standard

Synonym(s):

Lauric acid methyl ester, Methyl laurate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)10COOCH3
CAS Number:
Molecular Weight:
214.34
Beilstein:
1767780
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥99.5% (GC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

refractive index

n20/D 1.429-1.433

bp

261-262 °C (lit.)

mp

4.5-5 °C

density

0.869 g/mL at 20 °C (lit.)

format

neat

functional group

ester

shipped in

ambient

storage temp.

room temp

SMILES string

CCCCCCCCCCCC(=O)OC

InChI

1S/C13H26O2/c1-3-4-5-6-7-8-9-10-11-12-13(14)15-2/h3-12H2,1-2H3

InChI key

UQDUPQYQJKYHQI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Pictograms

Environment

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

282.2 °F - closed cup

Flash Point(C)

139 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

E Barboza-Silva et al.
Oral microbiology and immunology, 24(4), 265-271 (2009-07-04)
The arginine deiminase system (ADS) of oral bacteria is a major generator of alkali (ammonia) in dental plaque and is considered to have anticaries effects. However, many of the antimicrobial agents used in oral care products may reduce alkali production
D A Dimas et al.
AAPS PharmSciTech, 1(2), E16-E16 (2004-01-20)
The effects of coating thickness, type of adhesive, and type and concentration of enhancer on the mechanical properties of two acrylic pressure-sensitive adhesives (PSAs) were investigated using a 2(4) factorial design and an optimization technique. Sixteen formulations containing 0% or
Y Komata et al.
Biological & pharmaceutical bulletin, 17(5), 705-708 (1994-05-01)
The effect of long chain fatty acid (FA) and its analogs on the accumulation of thiamine disulfide (TDS) in rat skin using propylene glycol as a vehicle was studied in vitro. Lauric acid (12:0) increased the accumulation of TDS in
Jitender Sharma et al.
Journal of bioscience and bioengineering, 100(6), 662-666 (2006-02-14)
Efficient selective synthesis of the secondary amide surfactant N-methyl lauroylethanolamide from methyl laurate and N-methylethanol amine by carrier-fixed Chirazyme L-2 (Candida antarctica) using a kinetic strategy has been demonstrated. When different solvents were screened for product yields using Chirazyme L-2
Néstor M Carballeira et al.
Lipids, 40(10), 1063-1068 (2005-12-31)
The hitherto unknown 2-methylsulfanyldecanoic acid and 2-methylsulfanyldodecanoic acid were synthesized from methyl decanoate and methyl dodecanoate, respectively, through the reaction of lithium diisopropylamide and dimethyldisulfide in THF followed by saponification with potassium hydroxide in ethanol. Both alpha-methylsulfanylated FA were cytotoxic

Protocols

Protocol for GC Analysis of Bacterial Acid Methyl Esters (BAMEs) on Equity®-1

HPLC Analysis of Fatty Acid Methyl Esters (FAMES) on SUPELCOSIL™ LC-18

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service