Skip to Content
MilliporeSigma
All Photos(1)

Documents

900520

Sigma-Aldrich

2-Methyltetrahydrofuran

anhydrous, contains 250 ppm BHT as stabilizer, ZerO2®, ≥99.0%

Synonym(s):

2-Methyltetrahydrofuran ZerO2®, Degassed and low oxygen 2-methyltetrahydrofuran, Tetrahydrosilvan, 2-MeTHF, Tetrahydro-2-methylfuran

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H10O
CAS Number:
Molecular Weight:
86.13
Beilstein:
102448
EC Number:
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.07

grade

ZerO2®

Quality Level

Assay

≥99.0%

form

liquid

quality

anhydrous

contains

250 ppm BHT as stabilizer

refractive index

n20/D 1.406 (lit.)

bp

78-80 °C (lit.)

density

0.86 g/mL at 25 °C (lit.)

SMILES string

CC1CCCO1

InChI

1S/C5H10O/c1-5-3-2-4-6-5/h5H,2-4H2,1H3

InChI key

JWUJQDFVADABEY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

ZerO2 products are rigorously degassed with highly pure inert gas providing solvents and solutions (anhydrous if specified) with very low residual oxygen content.

Legal Information

ZerO2 is a registered trademark of Sigma-Aldrich Co. LLC

related product

Product No.
Description
Pricing

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Flam. Liq. 2 - Skin Irrit. 2

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

14.0 °F - closed cup

Flash Point(C)

-10.0 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service