Skip to Content
MilliporeSigma
2-Methyltetrahydrofuran anhydrous, contains 250 ppm BHT as stabilizer, ZerO2®, ≥99.0%

2-Methyltetrahydrofuran

2-Methyltetrahydrofuran anhydrous, contains 250 ppm BHT as stabilizer, ZerO2®, ≥99.0%
Synonym(s):
2-MeTHF, Tetrahydro-2-methylfuran, Tetrahydrosilvan
Empirical Formula (Hill Notation):
C5H10O
CAS Number:
Molecular Weight:
86.13
EC Number:

Products

Product Category
Brand
Sustainability
Boiling Point (°C)
Feature
Formula Weight
Physical Form
Product Line
Purity
Quality Segment
Markush Class
Markush Group
Special Grade
Technique