Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

89118

Supelco

Elemental Impurities according to ICH Q3D parenteral, Standard 1

(in 12% nitric acid), TraceCERT®, applicable for testing acc. to USP<232>, Ph.Eur. Gen. Chapter 5.20

Synonym(s):

ICH Q3D Elemental Impurity Analysis Standard

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material
TraceCERT®

Quality Level

Agency

according to ICH Q3D
according to Ph. Eur.
according to USP

product line

TraceCERT®

shelf life

limited shelf life, expiry date on the label

composition

Ag, 10 mg/L
As, 15 mg/L
Cd, 2 mg/L
Co, 5 mg/L
Hg, 3 mg/L
Ni, 20 mg/L
Pb, 5 mg/L
Se, 80 mg/L
Tl, 8 mg/L
V, 10 mg/L

concentration

(in 12% nitric acid)

technique(s)

ICP: suitable

format

multi-component solution

General description

This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to SI unit kg and measured against primary material from an NMI, e.g. NIST.
Certified content incl. uncertainty and expiry date are given on the certificate.
Download your certificate at: http://www.sigma-aldrich.com.

Application


  • Application of ICH Guidelines for the Assessment and Control of Elemental Impurities in Parecoxib Sodium by Graphite-Digestion and ICP-MS: This research demonstrates the application of ICH Q3D guidelines for evaluating elemental impurities in parenteral formulations, specifically using graphite-digestion and ICP-MS techniques on Parecoxib Sodium. This approach ensures compliance with pharmaceutical safety standards, emphasizing its critical role in maintaining drug quality and patient safety (Hao et al., 2022).

Preparation Note

For the preparation of this CRM only materials of highest purity were used.

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1B

Supplementary Hazards

WGK

WGK 2


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Control elemental impurities in drug products with analytical methods and materials to ensure patient safety.

Control elemental impurities in drug products with analytical methods and materials to ensure patient safety.

Control elemental impurities in drug products with analytical methods and materials to ensure patient safety.

Control elemental impurities in drug products with analytical methods and materials to ensure patient safety.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service