Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

84532

Sigma-Aldrich

Sarcosine

≥98.0% (T)

Synonym(s):

N-Methylglycine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3NHCH2COOH
CAS Number:
Molecular Weight:
89.09
Beilstein:
1699442
EC Number:
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:
NACRES:
NA.26
Pricing and availability is not currently available.

Product Name

Sarcosine, crystallized, ≥98.0% (T)

Quality Level

Assay

≥98.0% (T)

form

powder with small lumps

quality

crystallized

color

white

mp

208-212 °C (dec.) (lit.)
208-212 °C (dec.)

application(s)

cell analysis
peptide synthesis

SMILES string

CNCC(O)=O

InChI

1S/C3H7NO2/c1-4-2-3(5)6/h4H,2H2,1H3,(H,5,6)

InChI key

FSYKKLYZXJSNPZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Simone J P M Eussen et al.
The British journal of nutrition, 110(2), 363-374 (2012-12-12)
Different lifestyle patterns across Europe may influence plasma concentrations of B-vitamins and one-carbon metabolites and their relation to chronic disease. Comparison of published data on one-carbon metabolites in Western European regions is difficult due to differences in sampling procedures and
D S Lim et al.
Journal of the American College of Cardiology, 38(4), 1175-1180 (2001-10-05)
The goal of this study was to identify genes upregulated in the heart in human patients with hypertrophic cardiomyopathy (HCM). Hypertrophic cardiomyopathy is a genetic disease caused by mutations in contractile sarcomeric proteins. The molecular basis of diverse clinical and
Ryan O Walters et al.
Cell reports, 25(3), 663-676 (2018-10-18)
A hallmark of aging is a decline in metabolic homeostasis, which is attenuated by dietary restriction (DR). However, the interaction of aging and DR with the metabolome is not well understood. We report that DR is a stronger modulator of
Masato Hosokawa et al.
PloS one, 7(12), e52389-e52389 (2013-01-04)
In neurodegenerative disorders, abnormally hyperphosphorylated and aggregated tau accumulates intracellularly, a mechanism which is thought to induce neuronal cell death. Methylene blue, a type of phenothiazine, has been reported to inhibit tau aggregation in vitro. However, the effect of methylene
Eri Hara et al.
Biochimica et biophysica acta, 1830(8), 4046-4052 (2013-04-03)
Nanoparticle of Lactosome, which is composed of poly(l-lactic acid)-base depsipeptide with diameter of 35nm, accumulates in solid tumors by the enhanced permeability and retention (EPR) effect. However, a pharmacokinetic alteration of Lactosome was observed when Lactosome was repeatedly administered. This

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service