Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

83374

Sigma-Aldrich

Mercury(II) thiocyanate

purum p.a., ≥95.5% (complexometric)

Synonym(s):

Mercuric isothiocyanate, Mercuric sulfocyanide, Mercuric thiocyanate, Mercury di(thiocyanate), Thiocyanic acid, mercury(II) salt

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Hg(SCN)2
CAS Number:
Molecular Weight:
316.75
Beilstein:
3561344
EC Number:
MDL number:
UNSPSC Code:
12161600
PubChem Substance ID:
NACRES:
NA.22

grade

purum p.a.

Quality Level

Assay

≥95.5% (complexometric)

form

solid

reaction suitability

core: mercury
reagent type: catalyst

mp

165 °C (dec.) (lit.)

density

3.71 g/mL at 25 °C (lit.)

cation traces

Ca: ≤50 mg/kg
Cd: ≤50 mg/kg
Co: ≤50 mg/kg
Cu: ≤50 mg/kg
Fe: ≤50 mg/kg
K: ≤1000 mg/kg
Na: ≤100 mg/kg
Ni: ≤50 mg/kg
Pb: ≤50 mg/kg
Zn: ≤50 mg/kg

SMILES string

N#CS[Hg]SC#N

InChI

1S/2CHNS.Hg/c2*2-1-3;/h2*3H;/q;;+2/p-2

InChI key

GBZANUMDJPCQHY-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Application

Catalyst for addition of HSCN to alkynes.
Mercury(II) thiocyanate has been used for the determination of the chloride ions by colorimetric method.
It can be employed as a reactant for the synthesis of:
  • Polymeric mercury(II) coordination complexes with various derivatives of 4,4′-bipyridine ligands via the branched tube method.
  • Manganese mercury thiocyanate (MMTC), an organometallic optical single-crystal.
  • Heterotrinuclear copper(II)–mercury(II)–copper(II) complexes possessing antibacterial and antifungal activities.

Disclaimer

For U.S. Customers: Contains mercury; Do not place in trash - dispose according to local, state, or federal laws.

Signal Word

Danger

Hazard Classifications

Acute Tox. 1 Dermal - Acute Tox. 2 Inhalation - Acute Tox. 2 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - STOT RE 2

Supplementary Hazards

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

248.0 °F - closed cup

Flash Point(C)

120 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis, crystal structure and biological activity of two new heterotrinuclear thiocyanato bridged Cu (II)-Hg (II)-Cu (II) complexes.
Y?ld?r?m, LT, et al.
Polyhedron, 26(15), 4187-4194 (2007)
(4, 4′-Bipyridine) mercury (II) Coordination Polymers, Syntheses, and Structures.
Morsali A and Zhu LG
Helvetica Chimica Acta, 89(1), 81-93 (2006)
Surface modification of chromatography adsorbents by low temperature low pressure plasma.
Arpanaei A, et al.
Journal of Chromatography A, 1217(44), 6905-6916 (2010)
Barluenga, J.; Martinez-Gallo, J. M. et al.
J. Chem. Res. Synop., 274-274 (1986)
Crystal growth and characterization of the organometallic nonlinear optical crystal: manganese mercury thiocyanate (MMTC).
Wang X Q, et al.
Materials Research Bulletin, 36(5-6), 879-887 (2001)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service