Skip to Content
MilliporeSigma

1.04484

Mercury(II) thiocyanate

for analysis EMSURE® Reag. Ph Eur

Synonym(s):

Mercury(II) thiocyanate, Mercury(II) rhodanide, Mercury(II) thiocyanate

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
Hg(SCN)2
CAS Number:
Molecular Weight:
316.75
UNSPSC Code:
12352302
EC Index Number:
209-773-0
NACRES:
NA.21
MDL number:
Assay:
≥99.0% (complexometric)
Form:
solid
Solubility:
0.7 g/L
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


agency

reag. Ph. Eur.

Quality Segment

product line

EMSURE®

assay

≥99.0% (complexometric)

form

solid

potency

46 mg/kg LD50, oral (Rat)

ign. residue

≤0.05% (substances not reducing by formic acid)

pH

5 (20 °C, 50 g/L in H2O, slurry)

mp

165 °C (decomposition)

transition temp

flash point 120 °C

solubility

0.7 g/L

density

3.71 g/cm3 at 20 °C

bulk density

750 kg/m3

cation traces

Fe: ≤0.002%

storage temp.

2-30°C

SMILES string

[Hg+2].[K+].[K+].[S-]C#N.[S-]C#N.[S-]C#N.[S-]C#N

InChI

1S/4CHNS.Hg.2K/c4*2-1-3;;;/h4*3H;;;/q;;;;+2;2*+1/p-4

InChI key

HWXOCCNUUYRBKP-UHFFFAOYSA-J

Application



  • Spatial health risk assessments of nickel in the groundwater sources of a mining-impacted area: Mercury(II) thiocyanate may be used as a reagent in assessing heavy metal contamination, providing crucial data for environmental management and public health (Salehi et al., 2024).



Analysis Note

Assay (complexometric): ≥ 99.0 %
Fe (Iron): ≤ 0.002 %
Substances not reducing by formic acid (residue on ignition): ≤ 0.05 %
Corresponds to Reag. Ph Eur

Legal Information

EMSURE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

For U.S. Customers: Contains mercury; Do not place in trash - dispose according to local, state, or federal laws.


Still not finding the right product?

Explore all of our products under Mercury(II) thiocyanate


signalword

Danger

Hazard Classifications

Acute Tox. 1 Dermal - Acute Tox. 2 Inhalation - Acute Tox. 2 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - STOT RE 2

supp_hazards

Storage Class

6.1A - Combustible, acute toxic Cat. 1 and 2 / very toxic hazardous materials

wgk

WGK 3

flash_point_f

248.0 °F - closed cup

flash_point_c

120 °C - closed cup



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library