Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

78711

Sigma-Aldrich

RenLam® M-1

for microscopy, suitable as a embedding medium

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116121
NACRES:
MA.02

grade

for microscopy

Quality Level

form

liquid

refractive index

n/D 1.528-1.542

density

1.10-1.14 g/cm3 at 20 °C

application(s)

hematology
histology

storage temp.

room temp

Application

to be used with SIAL 69571 Ren HY 956 (Hardener for Epoxy Embedding Medium)

Legal Information

Renlam is a registered trademark of Huntsman Advanced Materials Inc.

related product

Product No.
Description
Pricing

replaced by

Product No.
Description
Pricing

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 2 - Eye Irrit. 2 - Skin Irrit. 2 - Skin Sens. 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 2


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service