Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

69571

Sigma-Aldrich

Ren HY 956

Epoxy Embedding Medium, Hardener

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12171500
NACRES:
MA.02

Quality Level

form

liquid

refractive index

n20/D 1.477-1.479

pH

12

density

1.005-1.009 g/mL at 20 °C

application(s)

hematology
histology

storage temp.

room temp

Application

to be used with SIAL 78711 RenLam M-1 (Epoxy Embedding Medium)

Legal Information

Ren is a trademark of Huntsman Advanced Materials GmbH

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 3 - Eye Dam. 1 - Skin Corr. 1B - Skin Sens. 1

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

305.6 °F - closed cup

Flash Point(C)

152 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service