Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

77970

Supelco

(S)-(−)-α-Methylbenzyl isocyanate

for chiral derivatization, LiChropur, ≥99.0%

Synonym(s):

(S)-(−)-1-Phenylethyl isocyanate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5CH(CH3)NCO
CAS Number:
Molecular Weight:
147.17
Beilstein:
4230972
EC Number:
MDL number:
UNSPSC Code:
12000000
PubChem Substance ID:
NACRES:
NA.22

grade

for chiral derivatization

Quality Level

Assay

≥99.0% (sum of enantiomers, GC)
≥99.0%

form

liquid

optical activity

[α]20/D −10.5±0.5°, neat

optical purity

enantiomeric ratio: ≥99.5:0.5 (GC)

quality

LiChropur

technique(s)

HPLC: suitable

refractive index

n20/D 1.513
n20/D 1.5145 (lit.)

bp

55-56 °C/2.5 mmHg (lit.)

density

1.045 g/mL at 20 °C (lit.)

storage temp.

2-8°C

SMILES string

C[C@H](N=C=O)c1ccccc1

InChI

1S/C9H9NO/c1-8(10-7-11)9-5-3-2-4-6-9/h2-6,8H,1H3/t8-/m0/s1

InChI key

JJSCUXAFAJEQGB-QMMMGPOBSA-N

Looking for similar products? Visit Product Comparison Guide

General description

(S)-(−)-α-Methylbenzyl isocyanate is a useful chiral derivatization reagent for all gas chromatography (GC) applications in the chiral field. It has been employed to meet the requirements for derivatization reagents for enantiomeric excess determinations.

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

LiChropur is a trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Classifications

Acute Tox. 2 Inhalation - Aquatic Chronic 3 - Eye Irrit. 2 - Resp. Sens. 1 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

149.0 °F - closed cup

Flash Point(C)

65 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Myriam Matoga et al.
Journal of enzyme inhibition and medicinal chemistry, 17(6), 375-379 (2003-04-10)
The derivatization of racemic 5-[(2-methylphenoxy)methyl]-2-amino-2-oxazoline, developed as an imidazoline binding sites ligand, with (+)-(R)-alpha-methylbenzyl isocyanate was performed in chloroform. The reaction led to two pairs of diastereomers, which were separated by RP-HPLC. A kinetic study of the derivatization reaction was
High-performance liquid chromatographic determination of the enantiomers of beta-adrenoceptor blocking agents in biological fluids. II. Studies with atenolol.
S K Chin et al.
Journal of chromatography, 489(2), 438-445 (1989-04-14)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service