Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

73074BC

Millipore

Assurance® GDS Vapor-Lock PCR Reagent

For use with Assurance® GDS

Synonym(s):

Mineral Oil for PCR Sample Prep

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116135
NACRES:
HA.03

Product Name

GDS Vapor-Lock, BioControl, For use with GDS

Quality Level

form

liquid

usage

sufficient for 30 tests

shelf life

1‑5 yr

packaging

vial of 1.2 mL

manufacturer/tradename

BioControl Systems 73074-30

storage condition

room temperature (Tightly closed in a dry, cool, and well-ventilated place)

technique(s)

PCR: suitable

color

colorless

application(s)

food and beverages
pathogen testing

storage temp.

room temp

General description

GDS Vapor-Lock (or mineral oil) is a hydrophobic, low-viscosity PCR encapsulation barrier. It is compatible with the Assurance® GDS Rotor-Gene® Q (RGQ) thermocycler.

Application

GDS Vapor-Lock is intended for use with positive control samples only. It minimizes the potential for the release of the amplified DNA into the laboratory environment.

Components

One vial, 1.3 mL, sufficient for 30 tests

Compatibility

BioControl GDS

Testing Method

PCR

Legal Information

Assurance is a registered trademark of Merck KGaA, Darmstadt, Germany
BioControl is a registered trademark of BioControl Systems, Inc.
Rotor-Gene is a registered trademark of Qiagen GmbH

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

437.0 °F - closed cup

Flash Point(C)

225.00 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service