Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

72795

Supelco

Butein

analytical standard

Synonym(s):

1-(2,4-Dihydroxyphenyl)-3-(3,4-dihydroxyphenyl)-2-propen-1-one, 2′,3,4,4′-Tetrahydroxychalcone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H12O5
CAS Number:
Molecular Weight:
272.25
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥98.0% (HPLC)

shelf life

limited shelf life, expiry date on the label

impurities

≤10% water (calc. from elemental analysis)

application(s)

food and beverages

format

neat

storage temp.

2-8°C

SMILES string

OC1=CC(O)=C(C(/C=C/C2=CC(O)=C(O)C=C2)=O)C=C1

InChI

1S/C15H12O5/c16-10-3-4-11(14(19)8-10)12(17)5-1-9-2-6-13(18)15(20)7-9/h1-8,16,18-20H/b5-1+

InChI key

AYMYWHCQALZEGT-ORCRQEGFSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Biochem/physiol Actions

Inhibits EGFR and Src tyrosine kinase activities; inhibits cAMP-dependent PDE-IV. Induces apoptosis in B16 melanoma cells and HL-60 human leukemia cells.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Rianne D Stowell et al.
Nature neuroscience, 22(11), 1782-1792 (2019-10-23)
Microglia are the brain's resident innate immune cells and also have a role in synaptic plasticity. Microglial processes continuously survey the brain parenchyma, interact with synaptic elements and maintain tissue homeostasis. However, the mechanisms that control surveillance and its role

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service