Skip to Content
MilliporeSigma

524488

Phloretin

A cell-permeable inhibtor of protein kinase C (IC50 = 20-50 µM).

Synonym(s):

Phloretin, 2ʹ,4ʹ,6ʹ-Trihydroxy-3- p-hydroxyphenylpropiophenone, 2ʹ,4ʹ,6ʹ-Trihydroxy-3-p-hydroxyphenylpropiophenone

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C15H14O5
CAS Number:
Molecular Weight:
274.27
UNSPSC Code:
12352200
NACRES:
NA.77
MDL number:
Assay:
≥97% (TLC)
Form:
solid
Storage condition:
OK to freeze
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

description

Merck USA index - 14, 7326

assay

≥97% (TLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

color

white

solubility

acetone: soluble, ethanol: soluble, methanol: soluble

shipped in

ambient

storage temp.

−20°C

SMILES string

Oc1c(c(cc(c1)O)O)C(=O)CCc2ccc(cc2)O

InChI

1S/C15H14O5/c16-10-4-1-9(2-5-10)3-6-12(18)15-13(19)7-11(17)8-14(15)20/h1-2,4-5,7-8,16-17,19-20H,3,6H2

InChI key

VGEREEWJJVICBM-UHFFFAOYSA-N

General description

A cell-permeable inhibtor of protein kinase C (IC50 = 20-50 µM). Activates Ca2+-activated K+ channels in amphibian myelinated nerve fibers at micromolar concentrations. An inhibitor of myo-inositol uptake. Also inhibits L-type Ca2+ channel activity. A potent inhibitor of 5′-iodothyronine deiodinase. Antagonist of prostaglandin F receptors linked to phospholipase C in astrocytes.

Biochem/physiol Actions

Product does not compete with ATP.
Reversible: no
Target IC50: 20-50 µM against protein kinase C
Cell permeable: yes

Preparation Note

Following reconstitution, aliquot and freeze (-20°C). Ethanol stock solutions are stable for up to 1 month at -20°C.

Other Notes

Berry, G.T., et al. 1994. Pediat. Res.35, 141.
Koh, D.S., et al. 1994. Neurosci. Lett. 165, 167.
Morreale de Escobar, G., et al. 1994. Endocrinology134, 2410.
Prevarskaya, N., et al. 1994. Mol. Cell. Neurosci.5, 699.
Kitanaka, J., et al. 1993. J. Neurochem.60, 704.
Wang, X., et al. 1993. Biochem. J.290, 249.
Benga, G., et al. 1992. Eur. J. Cell Biol.59, 219.
Jordan, M.L., and Wright, J. 1991. J. Surg. Res.50, 569.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Toxicity: Irritant (B)


Still not finding the right product?

Explore all of our products under Phloretin


pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

11 - Combustible Solids

wgk

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library