Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

67253

Supelco

Sodium formate

analytical standard

Synonym(s):

E237, Formic acid sodium salt

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HCOONa
CAS Number:
Molecular Weight:
68.01
Beilstein:
3595134
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

99.0-101.0% (wt., NT)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

pH

7.0-8.5

mp

259-262 °C (lit.)

solubility

H2O: 10%, clear, colorless

UV absorption

λ: 260 nm Amax: ≤0.05
λ: 270 nm Amax: ≤0.04
λ: 300 nm Amax: ≤0.03
λ: 330 nm Amax: ≤0.02
λ: 500 nm Amax: ≤0.02

application(s)

cleaning products
cosmetics
food and beverages
personal care

format

neat

SMILES string

[Na+].[O-]C=O

InChI

1S/CH2O2.Na/c2-1-3;/h1H,(H,2,3);/q;+1/p-1

InChI key

HLBBKKJFGFRGMU-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Pravin K Mishra et al.
Journal of neurochemistry, 146(6), 722-734 (2018-07-03)
Depression is one of the most debilitating neuropsychiatric disorders. Most of the current antidepressants have long remission time and low recovery rate. This study explores the impact of ketamine on neuronal and astroglial metabolic activity in prefrontal cortex in a
Sergey Zakharov et al.
Kidney international, 86(1), 199-207 (2014-03-14)
During an outbreak of methanol poisonings in the Czech Republic in 2012, we were able to study methanol and formate elimination half-lives during intermittent hemodialysis (IHD) and continuous veno-venous hemodialysis/hemodiafiltration (CVVHD/HDF) and the relative impact of dialysate and blood flow
Sergey Zakharov et al.
Basic & clinical pharmacology & toxicology, 116(5), 445-451 (2014-10-14)
The aim of this article was to study the role of serum formate (S-formate) in diagnosing methanol poisoning. A prospective study was undertaken of 38 patients from the Czech methanol mass poisoning in 2012 - median age 51 [interquartile range
Sergey Zakharov et al.
Biomedical papers of the Medical Faculty of the University Palacky, Olomouc, Czechoslovakia, 158(4), 641-649 (2014-12-09)
During an outbreak of mass methanol poisonings in the Czech Republic in 2012-2013, fomepizole was applied as an alternative antidote to ethanol. We present the laboratory data, clinical features, adverse reactions, and treatment outcomes in all patients treated with fomepizole.
Susanta Mondal et al.
PloS one, 10(1), e0116566-e0116566 (2015-01-09)
Upregulation and/or maintenance of regulatory T cells (Tregs) during an autoimmune insult may have therapeutic efficacy in autoimmune diseases. Although several immunomodulatory drugs and molecules are available, most present significant side effects over long-term use. Cinnamon is a commonly used

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service