Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

63936

Sigma-Aldrich

Ethyl O-(2-mesitylenesulfonyl)acethydroxamate

purum, ≥97.0% (HPLC)

Synonym(s):

O-(2-Mesitylenesulfonyl)acethydroxamic acid ethyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
2,4,6-(CH3)3C6H2SO2ON=C(CH3)OC2H5
CAS Number:
Molecular Weight:
285.36
Beilstein:
2144156
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

grade

purum

Quality Level

Assay

≥97.0% (HPLC)

form

solid

mp

54-56 °C (lit.)
54-58 °C

functional group

amine
sulfonic acid

storage temp.

2-8°C

SMILES string

CCO\C(C)=N\OS(=O)(=O)c1c(C)cc(C)cc1C

InChI

1S/C13H19NO4S/c1-6-17-12(5)14-18-19(15,16)13-10(3)7-9(2)8-11(13)4/h7-8H,6H2,1-5H3/b14-12+

InChI key

KQCBSWBQAXTILK-WYMLVPIESA-N

Looking for similar products? Visit Product Comparison Guide

Other Notes

Stable precursor of O-mesitylenesulfonylhydroxylamine (MSH) — a powerful aminating reagent; review

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Y. Taura et al.
Synthesis, 1-1 (1977)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service