Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

213136

Sigma-Aldrich

Hydroxylamine-O-sulfonic acid

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H2NOSO3H
CAS Number:
Molecular Weight:
113.09
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
NACRES:
NA.21

Quality Level

Assay

97%

mp

210 °C (dec.) (lit.)

storage temp.

2-8°C

SMILES string

NOS(O)(=O)=O

InChI

1S/H3NO4S/c1-5-6(2,3)4/h1H2,(H,2,3,4)

InChI key

DQPBABKTKYNPMH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Hydroxylamine-O-sulfonic acid is a versatile synthetic reagent. It is widely employed as a nucleophile and as an electrophile in various organic syntheses. It can be synthesized by reacting hydroxylamine sulfate with 30% fuming H2SO4.

Application

Hydroxylamine-O-sulfonic acid (HOSA) may be employed in the preparation of the following:
  • monosubstituted and 1,1-disubstituted hydrazines
  • symmetrically substituted pyrroles
  • aniline
  • C-substituted amino derivatives
  • lactams
Hydroxylamine-O-sulfonic acid may be used in the following processes:
As a nitrogen transfer reagent for the conversion of aldehydes to nitriles in aqueous medium.
Synthesis of N-aminopiperidine (NAPP) by reacting with piperidine.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 3 - Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

A New Strategy for the Preparation of N-Aminopiperidine Using Hydroxylamine-O-Sulfonic Acid: Synthesis, Kinetic Modelling, Phase Equilibria, Extraction and Processes."
Labarthe E, et al.
Advances in Chemical Engineering and Science, 3(2) (2013)
Direct synthesis of nitriles from aldehydes with hydroxylamine-O-sulfonic acid in acidic water.
Quinn DJ, et al.
Tetrahedron Letters, 57(34), 3844-3847 (2016)
Hydroxylamine-O-sulfonic Acid.
Erdik E and Saczewski J.
e-EROS Encyclopedia of Reagents for Organic Synthesis. (2013)
Synthetic Methods and Reactions; 671. One-Step Conversion of Alicyclic Ketones into Lactams with Hydroxylamine-O-sulfonic Acid/Formic Acid.
Olah GA and Fung AP.
Synthesis, 07, 537-538 (1979)
K Kohda et al.
Nucleic acids symposium series, (17)(17), 145-148 (1986-01-01)
Amination of guanosine (Guo) with 2,4-dinitrophenoxyamine in aqueous DMF gave 7-amino-Guo, which was readily converted to 8,5'-O-cyclo-Guo, and 8-hydroxy-Guo. Deoxyguanosine (dG) gave only deglycosylated 7-amino-G under the same reaction condition. Aminations of Guo and dG with hydroxylamine-O-sulfonic acid above pH

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service