Skip to Content
MilliporeSigma
All Photos(1)

Documents

63237

Millipore

TB-Medium Base according to Loewenstein-Jensen

suitable for microbiology, NutriSelect® Plus

Synonym(s):

Loewenstein-Jensen Medium (Base)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41171607
NACRES:
NA.85

sterility

non-sterile

Quality Level

form

powder

shelf life

limited shelf life, expiry date on the label

composition

L-asparagine, 2.25 g/L
magnesium citrate, 0.375 g/L
magnesium sulfate, 0.15 g/L
malachite green, 0.25 g/L
monopotassium phosphate, 1.5 g/L
potato flour, 18.75 g/L

manufacturer/tradename

NutriSelect® Plus

technique(s)

microbiological culture: suitable

application(s)

clinical testing
environmental
food and beverages
life science and biopharma
veterinary

microbiology

suitability

mycobacteria

Application

The selective Lowenstein Jensen Medium is used for isolation of Mycobacteria sp. from contaminated specimens. Egg supplement enhances Mycobacteria growth. Malachite green, penicillin* and nalidixic acid* inhibit Gram-negative and Gram-positive bacteria. (* included in the Gruft Supplement)

Preparation Note

Suspend 37,2 g in 600 ml of purified water containing 12 ml of glycerol (Cat. No. 49769). Heat with frequent agitation and boil for one minute. Sterilize at 121°C for 15 minutes. Cool to 45-50°C and, aseptically, add 1 L of homogenised sterile eggs and if required 4 vials of Gruft Mycobacterial Supplement (Cat. No. 51803). Dispense into sterile culture tubes and coagulate at 85°C for 45 minutes in slant position.

Footnote

We offer two media types: the superior granulated GranuCult® and the cost-efficient powdered NutriSelect® culture media, depending on your needs.
The designations basic, plus, or prime are added to indicate the quality control level, from basic quality control to standard QC plus to prime for full regulatory compliance.

Legal Information

GRANUCULT is a registered trademark of Merck KGaA, Darmstadt, Germany
NutriSelect is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Identifying Mycobacterium species crucial due to varying clinical significance; some pathogenic, others not.

Identifying Mycobacterium species crucial due to varying clinical significance; some pathogenic, others not.

Identifying Mycobacterium species crucial due to varying clinical significance; some pathogenic, others not.

Identifying Mycobacterium species crucial due to varying clinical significance; some pathogenic, others not.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service