Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

63036

Sigma-Aldrich

Magnesium chloride solution

BioUltra, for molecular biology, ~0.025 M in H2O

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
MgCl2
CAS Number:
Molecular Weight:
95.21
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.26

grade

for molecular biology

Quality Level

product line

BioUltra

concentration

~0.025 M in H2O

impurities

DNases, none detected
RNases, none detected
insoluble matter, passes filter test
phosphatases, none detected
proteases, none detected
≤0.0002% total nitrogen (N)

pH

5.0-7.0 (25 °C)

anion traces

phosphate (PO43-): ≤5 mg/kg
sulfate (SO42-): ≤20 mg/kg

cation traces

Al: ≤1 mg/kg
As: ≤0.1 mg/kg
Ba: ≤1 mg/kg
Bi: ≤1 mg/kg
Ca: ≤10 mg/kg
Cd: ≤1 mg/kg
Co: ≤1 mg/kg
Cr: ≤1 mg/kg
Cu: ≤1 mg/kg
Fe: ≤1 mg/kg
K: ≤500 mg/kg
Li: ≤1 mg/kg
Mn: ≤1 mg/kg
Mo: ≤1 mg/kg
Na: ≤1000 mg/kg
Ni: ≤1 mg/kg
Pb: ≤1 mg/kg
Sr: ≤1 mg/kg
Zn: ≤1 mg/kg

λ

neat

UV absorption

λ: 260 nm Amax: 0.03
λ: 280 nm Amax: 0.03

SMILES string

Cl[Mg]Cl

InChI

1S/2ClH.Mg/h2*1H;/q;;+2/p-2

InChI key

TWRXJAOTZQYOKJ-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Packaging

package of 10 × 1 mL ampouls

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Stephen Q Wong et al.
Oncotarget, 6(2), 1115-1127 (2014-12-30)
Melanoma is often caused by mutations due to exposure to ultraviolet radiation. This study reports a recurrent somatic C > T change causing a P131L mutation in the RQCD1 (Required for Cell Differentiation1 Homolog) gene identified through whole exome sequencing
Hideyuki Arita et al.
Brain tumor pathology, 32(1), 22-30 (2014-04-22)
Assessment of the mutational status of the isocitrate dehydrogenase 1/2 (IDH1/2) gene has become an integral part of the standard diagnostic procedure and, therefore, needs to be accurate. This may, however, be compromised by various factors including the method of
Elaine T Lim et al.
PLoS genetics, 10(7), e1004494-e1004494 (2014-08-01)
Exome sequencing studies in complex diseases are challenged by the allelic heterogeneity, large number and modest effect sizes of associated variants on disease risk and the presence of large numbers of neutral variants, even in phenotypically relevant genes. Isolated populations
Bert De Rybel et al.
Science (New York, N.Y.), 345(6197), 1255215-1255215 (2014-08-12)
Coordination of cell division and pattern formation is central to tissue and organ development, particularly in plants where walls prevent cell migration. Auxin and cytokinin are both critical for division and patterning, but it is unknown how these hormones converge
Manal A Farg et al.
Human molecular genetics, 23(13), 3579-3595 (2014-02-20)
Intronic expansion of a hexanucleotide GGGGCC repeat in the chromosome 9 open reading frame 72 (C9ORF72) gene is the major cause of familial amyotrophic lateral sclerosis (ALS) and frontotemporal dementia. However, the cellular function of the C9ORF72 protein remains unknown.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service