Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

60322

Supelco

Potassium hexafluorosilicate

≥99.0% (T)

Synonym(s):

Dipotassium hexafluorosilicate, Potassium fluorosilicate, Potassium fluosilicate, Potassium silicofluoride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
K2SiF6
CAS Number:
Molecular Weight:
220.27
EC Number:
MDL number:
UNSPSC Code:
12000000
PubChem Substance ID:
NACRES:
NA.21
Assay:
≥99.0% (T)
form:
solid

Quality Level

Assay

≥99.0% (T)

form

solid

density

2.665 g/mL at 25 °C (lit.)

anion traces

chloride (Cl-): ≤500 mg/kg
sulfate (SO42-): ≤200 mg/kg

cation traces

Ca: ≤50 mg/kg
Cd: ≤50 mg/kg
Co: ≤50 mg/kg
Cu: ≤50 mg/kg
Fe: ≤100 mg/kg
Na: ≤1000 mg/kg
Ni: ≤50 mg/kg
Pb: ≤50 mg/kg
Zn: ≤50 mg/kg

SMILES string

[K+].[K+].F[Si--](F)(F)(F)(F)F

InChI

1S/F6Si.2K/c1-7(2,3,4,5)6;;/q-2;2*+1

InChI key

UUHPPUWEDCVPCO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Caution

may contain black particles

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ali Ouasri et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 59(4), 851-857 (2003-03-01)
The X-ray powder diffraction pattern of [(CH(3))(3)NH](2)SiF(6) was obtained and indexed on the basis of a centred cubic unit cell with the P4(1)32 as the likely space group. The Infrared and Raman spectra of this compound have been recorded at
Shingo Shibata et al.
American journal of dentistry, 24(3), 148-152 (2011-08-31)
To evaluate the degree of penetration of an ammonium hexafluorosilicate [SiF: (NH4)2SiF6] solution containing various antibacterial agents into dentin and the depth of dentin tubule occlusion by the precipitate. Various antibacterial SiF solutions were prepared with the addition to chlorhexidine
Ram Thaimattam et al.
Acta crystallographica. Section B, Structural science, 64(Pt 4), 483-490 (2008-07-22)
The co-crystal structure of N-methylpiperidine betaine with N-methylpiperidine betaine hexafluorosilicate represents an unusual case of a salt co-crystal with a high Z' value (3), unexpected conformational variability, and with nearly 50% of its contents disordered. The betaine units from the
Si Yang et al.
Analytical chemistry, 81(9), 3478-3484 (2009-04-10)
We developed an alcohol-free sol-gel approach to encapsulate biomolecules such as horseradish peroxidase (HRP) in an electrochemically induced three-dimensional porous silica matrix by a one-step process. In this sol-gel process, the electrochemically generated hydroxyl ions at the electrode surface by
Mark A Brzezinski et al.
Analytical chemistry, 78(17), 6109-6114 (2006-09-02)
A procedure for the automated determination of isotopic abundances of silicon from biogenic and lithogenic particulate matter and from dissolved silicon in fresh or saltwaters is reported. Samples are purified using proven procedures through the reaction of Si with acidified

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service