Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

556505

Sigma-Aldrich

4-Chloro-7-chlorosulfonyl-2,1,3-benzoxadiazole

97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H2Cl2N2O3S
CAS Number:
Molecular Weight:
253.06
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:
NACRES:
NA.47

Quality Level

Assay

97%

form

powder or crystals

mp

86-90 °C (lit.)

λmax

324 nm

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

room temp

SMILES string

Clc1ccc(c2nonc12)S(Cl)(=O)=O

InChI

1S/C6H2Cl2N2O3S/c7-3-1-2-4(14(8,11)12)6-5(3)9-13-10-6/h1-2H

InChI key

BPPRLMZEVZSIIJ-UHFFFAOYSA-N

Application

Bifunctional fluorescent reagent. Cross-linking reagent

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Graphene is the building block for carbon nanomaterials with different dimensionalities.

Graphene is the building block for carbon nanomaterials with different dimensionalities.

Graphene is the building block for carbon nanomaterials with different dimensionalities.

Graphene is the building block for carbon nanomaterials with different dimensionalities.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service