Skip to Content
MilliporeSigma
All Photos(1)

Documents

54362

Supelco

ICP solvent

for inorganic trace analysis, ≥99.9995% (metals basis)

Synonym(s):

Hydrotreated light distillate (petroleum)

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
EC Number:
UNSPSC Code:
12000000
NACRES:
NA.24

grade

for inorganic trace analysis

Quality Level

Assay

≥99.9995% (metals basis)

technique(s)

AAS: suitable
ICP: suitable

bp

200-250 °C (lit.)

mp

-58 °C (lit.)

density

0.810-0.838 g/mL at 20 °C

anion traces

phosphate (PO43-): ≤1 mg/kg
sulfate (SO42-): ≤5 mg/kg

cation traces

Ag: ≤1 μg/kg
Al: ≤1 μg/kg
As: ≤1 μg/kg
Au: ≤1 μg/kg
Ba: ≤0.5 μg/kg
Be: ≤0.5 μg/kg
Bi: ≤0.5 μg/kg
Ca: ≤20 μg/kg
Cd: ≤0.5 μg/kg
Ce: ≤0.5 μg/kg
Co: ≤0.5 μg/kg
Cr: ≤1 μg/kg
Cu: ≤1 μg/kg
Fe: ≤5 μg/kg
Ga: ≤0.5 μg/kg
Ge: ≤0.5 μg/kg
In: ≤0.5 μg/kg
Ir: ≤0.5 μg/kg
K: ≤10 μg/kg
Li: ≤0.5 μg/kg
Mg: ≤1 μg/kg
Mn: ≤0.5 μg/kg
Mo: ≤0.5 μg/kg
Na: ≤10 μg/kg
Ni: ≤1 μg/kg
Pb: ≤0.5 μg/kg
Pd: ≤1 μg/kg
Pt: ≤0.5 μg/kg
Rb: ≤0.5 μg/kg
Rh: ≤0.5 μg/kg
Ru: ≤0.5 μg/kg
Sb: ≤0.5 μg/kg
Sn: ≤0.5 μg/kg
Sr: ≤0.5 μg/kg
Ti: ≤1 μg/kg
Tl: ≤0.5 μg/kg
U: ≤0.5 μg/kg
V: ≤0.5 μg/kg
Zn: ≤50 μg/kg
Zr: ≤0.5 μg/kg

Application

ICP solvent is low odor and reduced toxicity equivalent to classical kerosene or xylene and ideal for dilution of oils and other organic fluids for determination of metal traces.

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Asp. Tox. 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

158.0 °F

Flash Point(C)

70 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Fabrício E Bortot Coelho et al.
Journal of environmental science and health. Part A, Toxic/hazardous substances & environmental engineering, 56(3), 289-302 (2021-01-15)
Galvanizing industries generate large amounts of effluents rich in toxic and carcinogenic chromium(VI) species. Effective and sustainable treatments are required to comply with environmental regulations. This work focused on the development of innovative treatments for Cr(VI) by its removal from
Venita Totten et al.
Forensic science international, 312, 110309-110309 (2020-05-14)
Significant amounts of water are used to extinguish fires, and finding evidence of ignitable liquid residue can be challenging for investigators. Hydrophobic pads have been designed to collect oil-based products from the surface of water and preferentially absorb non-polar compounds

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service