Skip to Content
MilliporeSigma
All Photos(1)

Documents

53638

Supelco

Nitrate Nitrogen Standard for IC

certified reference material, 1000 mg/L N as nitrate in water (nominal concentration), TraceCERT®

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material
TraceCERT®

Quality Level

product line

TraceCERT®

concentration

1000 mg/L N as nitrate in water (nominal concentration)

technique(s)

ion chromatography: suitable

application(s)

environmental
food and beverages

format

single component solution

General description

This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to SI unit kg and measured against primary material from an NMI, e.g. NIST.
Certified content incl. uncertainty and expiry date are given on the certificate.
Download your certificate at: http://www.sigma-aldrich.com.

Application

<ul>
<li>
<strong>Anaerobic Ammonium Oxidation (Anammox) with Planktonic Cells in a Redox-Stable Semicontinuous Stirred-Tank Reactor:</strong> This study explores the use of Anammox processes for efficient nitrogen removal from wastewater, underlining the importance of precise nitrate nitrogen standards for ensuring accurate process control and environmental compliance (Ding et al., 2018).
</li>
<li>
<strong>Enhancement of total lipid yield by nitrogen, carbon, and iron supplementation in isolated microalgae:</strong> The research demonstrates how nutrient management, including controlled nitrate nitrogen levels, can significantly impact lipid production in algae, vital for biofuel development and other biotechnological applications (Sivaramakrishnan and Incharoensakdi, 2017).
</li>
<li>
<strong>Seasonal trends of atmospheric nitrogen dioxide and sulfur dioxide over North Santa Clara, Cuba:</strong> This environmental monitoring study highlights the necessity of accurate calibration standards like nitrate nitrogen for assessing air quality and studying atmospheric chemistry (Alejo et al., 2013).
</li>
<li>
<strong>Analysis of inorganic nitrogen and related anions in high salinity water using ion chromatography:</strong> The paper details methodologies that incorporate nitrate nitrogen standards to ensure the reliability of ion chromatography results, crucial for environmental monitoring and regulatory compliance (Wilson et al., 2011).
</li>
<li>
<strong>Simultaneous determination of bromide, nitrite and nitrate ions in seawater by capillary zone electrophoresis:</strong> This research uses nitrate nitrogen standards to calibrate the analytical methods needed for precise quantification of nutrients in marine environments, aiding in ecological assessments and marine chemistry (Fukushi et al., 2000).
</li>
</ul>

Linkage

Visit the IC Portal to learn more

Analysis Note

1000 mg/L N as nitrate in water, prepared with high purity NaNO3 and water

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

related product

Product No.
Description
Pricing

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service