Skip to Content
MilliporeSigma
All Photos(4)

Key Documents

493538

Sigma-Aldrich

Ethyl alcohol, Pure

greener alternative

94.9-96.0%, 190 proof, meets USP testing specifications, USP/NF

Synonym(s):

Ethanol, absolute alcohol, non-denatured ethanol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH2OH
CAS Number:
Molecular Weight:
46.07
EC Number:
MDL number:
UNSPSC Code:
15101802
PubChem Substance ID:
NACRES:
NA.21
Assay:
94.9-96.0%
bp:
78.3 °C

Product Name

Ethyl alcohol, Pure, 190 proof, meets USP testing specifications

Agency

USP/NF
meets USP testing specifications

Quality Level

vapor density

1.59 (vs air)

Assay

94.9-96.0%

form

liquid

expl. lim.

3.1-27.7 %

availability

not available in (Sales restictions may apply)

greener alternative product characteristics

Safer Solvents and Auxiliaries
Use of Renewable Feedstocks
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

concentration

190 proof

evapn. residue

≤0.0025%

color

colorless

pH

7 (20 °C, 10 g/L)

bp

78.3 °C

mp

-114 °C

solubility

water: soluble

λ

in H2O

UV absorption

λ: 240 nm Amax: 0.40
λ: 250 nm Amax: 0.30
λ: 260 nm Amax: 0.30
λ: 270 nm Amax: 0.10
λ: 340 nm Amax: 0.1

greener alternative category

SMILES string

OCC

InChI

1S/C2H6O/c1-2-3/h3H,2H2,1H3

InChI key

LFQSCWFLJHTTHZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Prices include Federal Tax.
The product is a non-denatured 190proof ethanol. Ethanol, a primary alcohol, is an important raw material for the synthesis of various compounds. It can be synthesized either from molasses by fermentation or by hydration of ethylene. It acts as a hydrogen source and reaction medium during the transfer hydrogenation of carbonyl compounds in the presence of sodium hydroxide. The impact of ethanol/water fumigation on the performance of diesel engines has been studied.
We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. Ethanol is environmentally preferable green solvent and thus it aligns with "Safer Solvents and Auxiliaries" and "Use of Renewable Feedstocks". 

Application


  • Improved isoprene detection performance of Si-doped WO(3) films deposited by sputtering and post-annealing: Highlights the application of pure ethyl alcohol as a solvent in the deposition process for sensors, enhancing isoprene detection capabilities, vital for environmental monitoring and industrial safety (Lin et al., 2024).

  • An efficient method for the preparative isolation and purification of alkaloids from Gelsemium by using high speed counter-current chromatography and preparative HPLC: Demonstrates the use of pure ethyl alcohol in high-performance liquid chromatography, critical for the extraction and purification of bioactive compounds in pharmaceutical research (Liang et al., 2024).

  • Aging processes in dental thermoplastics - Thermoanalytical investigations and effects on Vickers as well as Martens hardness: Utilizes pure ethyl alcohol in the processing of dental materials to assess their durability and physical properties, emphasizing its role in material science and dentistry (Kreitczick et al., 2024).

  • Functionalization of bamboo fibers with lawsone dye (Lawsonia inermis) to produce bioinspired hybrid color composite with antibacterial activity: Features the application of pure ethyl alcohol in the treatment of bamboo fibers, contributing to the development of functional materials with enhanced properties (Szadkowski et al., 2024).

  • Facile synthesis of Ag lattice doped mesoporous In(2)O(3) nanocubes for high performance ethanol sensing: Uses pure ethyl alcohol in the synthesis process of nanocubes for enhanced ethanol sensing capabilities, pivotal for developing sensitive and efficient sensors (Liu et al., 2024).

Other Notes

Sales restrictions may apply

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

55.4 °F - closed cup

Flash Point(C)

13 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Assessment of a direct injection diesel engine fumigated with ethanol/water mixtures.
Morsy MH.
Energy Conversion and Management , 94, 406-414 (2015)
Sodium hydroxide-catalyzed transfer hydrogenation of carbonyl compounds and nitroarenes using ethanol or isopropanol as both solvent and hydrogen donor.
Wang D, et al.
J. Mol. Catal. A: Chem., 400, 14-21 (2015)
Ross B Mounsey et al.
Experimental neurology, 273, 36-44 (2015-08-06)
Parkinson's disease (PD) is a common chronic neurodegenerative disorder, usually of idiopathic origin. Symptoms including tremor, bradykinesia, rigidity and postural instability are caused by the progressive loss of dopaminergic neurons in the nigrostriatal region of the brain. Symptomatic therapies are
Cem Kuscu et al.
Nature biotechnology, 32(7), 677-683 (2014-05-20)
RNA-guided genome editing with the CRISPR-Cas9 system has great potential for basic and clinical research, but the determinants of targeting specificity and the extent of off-target cleavage remain insufficiently understood. Using chromatin immunoprecipitation and high-throughput sequencing (ChIP-seq), we mapped genome-wide
Gian Marco Leggio et al.
Neuropsychopharmacology : official publication of the American College of Neuropsychopharmacology, 39(8), 2017-2028 (2014-03-04)
Mesolimbic dopamine (DA) controls drug- and alcohol-seeking behavior, but the role of specific DA receptor subtypes is unclear. We tested the hypothesis that D3R gene deletion or the D3R pharmacological blockade inhibits ethanol preference in mice. D3R-deficient mice (D3R(-/-)) and

Protocols

To measure alcohol dehydrogenase activity, this assay uses β-nicotinamide adenine dinucleotide phosphate and a continuous spectrophotometric rate determination at 340 nm.

Related Content

Why should you have to choose between solvents that are ecological and those that are reliable? Enjoy both at once with our biorenewable and greener solutions. Cyrene™ solvent is a new dipolar aprotic alternative to common REACH restricted solvents, such as N methyl-2-pyrrolidone (NMP) and Dimethylformamide (DMF).

Why should you have to choose between solvents that are ecological and those that are reliable? Enjoy both at once with our biorenewable and greener solutions. Cyrene™ solvent is a new dipolar aprotic alternative to common REACH restricted solvents, such as N methyl-2-pyrrolidone (NMP) and Dimethylformamide (DMF).

Why should you have to choose between solvents that are ecological and those that are reliable? Enjoy both at once with our biorenewable and greener solutions. Cyrene™ solvent is a new dipolar aprotic alternative to common REACH restricted solvents, such as N methyl-2-pyrrolidone (NMP) and Dimethylformamide (DMF).

Why should you have to choose between solvents that are ecological and those that are reliable? Enjoy both at once with our biorenewable and greener solutions. Cyrene™ solvent is a new dipolar aprotic alternative to common REACH restricted solvents, such as N methyl-2-pyrrolidone (NMP) and Dimethylformamide (DMF).

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service