Skip to Content
MilliporeSigma
All Photos(3)

Documents

465666

Sigma-Aldrich

Magnesium

ReagentPlus®, powder, −325 mesh, 99.5% trace metals basis

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Mg
CAS Number:
Molecular Weight:
24.31
Beilstein:
4948473
EC Number:
MDL number:
UNSPSC Code:
12141502
PubChem Substance ID:
NACRES:
NA.22

vapor pressure

1 mmHg ( 621 °C)

Quality Level

description

Metals basis purity

product line

ReagentPlus®

Assay

99.5% trace metals basis

form

powder

autoignition temp.

950 °F

reaction suitability

reagent type: reductant

resistivity

4.46 μΩ-cm, 20°C

particle size

−325 mesh

bp

1090 °C (lit.)

mp

648 °C (lit.)

density

1.74 g/mL at 25 °C (lit.)

SMILES string

[Mg]

InChI

1S/Mg

InChI key

FYYHWMGAXLPEAU-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Magnesium is generally used in the preparation of various organomagnesium compounds such as Grignard reagents, that are widely used in organic synthesis.
It can also be used to catalyze Pinacol coupling, Reformatsky reaction and degradation of carcinogenic polycyclic aromatic hydrocarbons (PAHs).

Legal Information

ReagentPlus is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Flame

Signal Word

Danger

Hazard Statements

Hazard Classifications

Pyr. Sol. 1 - Water-react 1

Storage Class Code

4.2 - Pyrophoric and self-heating hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The Grignard Reagents.
Seyferth D
Organometallics, 28 (6), 1598-1605 (2009)
The quest for chiral Grignard reagents.
Hoffmann RW
Chemical Society Reviews, 32(4), 225-230 (2003)
Magnesium-induced pinacol coupling of aromatic aldehydes and ketones in water under ultrasound.
Li JT, et al.
Indian Journal of Chemistry, 43B, 196-198 (2004)
Magnesium: Annual survey covering the year 1971.
Blomberg C
Journal of Organometallic Chemistry, 45, 1-137 (1972)
Optimizing Reductive Degradation of PAHs Using Anhydrous Ethanol with Magnesium Catalyzed by Glacial Acetic Acid.
Al Shra?ah A, et al.
ACS Omega, 3(3), 3554-3561 (2018)

Articles

Mechanochemical Effect of Severe Plastic Deformations: Metal Alloys, Hydrides and Molecular Solids

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service