Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

8.18506

Sigma-Aldrich

Magnesium

powder (particle size < 0.1 mm) for synthesis

Synonym(s):

Magnesium

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Mg
CAS Number:
Molecular Weight:
24.31
MDL number:
UNSPSC Code:
12141502
EC Index Number:
231-104-6
NACRES:
NA.22

vapor pressure

0.00013 hPa ( 325 °C)

Quality Level

form

solid

autoignition temp.

>450 °C

bp

1107 °C/1013 hPa

mp

651 °C

density

1.74 g/cm3 at 20 °C

storage temp.

2-30°C

SMILES string

[Mg+2]

InChI

1S/Mg/q+2

InChI key

JLVVSXFLKOJNIY-UHFFFAOYSA-N

Analysis Note

Assay (on Mg): ≥ 97.0 %

Pictograms

Flame

Signal Word

Danger

Hazard Statements

Hazard Classifications

Pyr. Sol. 1 - Water-react 1

Storage Class Code

4.2 - Pyrophoric and self-heating hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service