Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

45398

Supelco

Chlorthiamid

PESTANAL®, analytical standard

Synonym(s):

2,6-Dichlorobenzenecarbothioamide, 2,6-Dichlorothiobenzamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H5Cl2NS
CAS Number:
Molecular Weight:
206.09
Beilstein:
1910353
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

SMILES string

NC(=S)c1c(Cl)cccc1Cl

InChI

1S/C7H5Cl2NS/c8-4-2-1-3-5(9)6(4)7(10)11/h1-3H,(H2,10,11)

InChI key

KGKGSIUWJCAFPX-UHFFFAOYSA-N

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

>212.0 °F

Flash Point(C)

> 100 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

L Bruun et al.
Journal of immunological methods, 240(1-2), 133-142 (2000-06-16)
2,6-Dichlorobenzamide (BAM) is the dominant degradation product in soil of the widely used herbicide dichlobenil. To detect BAM in water, a highly sensitive and specific enzyme-linked immunosorbent assay (ELISA) was developed. As an alternative to conventional coating of ELISA plates
J E Bakke et al.
Xenobiotica; the fate of foreign compounds in biological systems, 18(9), 1063-1075 (1988-09-01)
1. Twelve 14C-labelled metabolites were isolated from either urine or bile from either rats (11 metabolites) or goats (7 metabolites) given single oral doses of 2,6-dichlorobenzo[14C]nitrile (DCBN). Five of these metabolites were also excreted in urine from rats dosed orally
E B Brittebo et al.
Fundamental and applied toxicology : official journal of the Society of Toxicology, 17(1), 92-102 (1991-07-01)
The toxic effects of the herbicide chlorthiamid (2,6-dichlorothiobenzamide) and its major environmental metabolite 2,6-dichlorobenzamide (DCBA) were examined in the nasal passages of C57Bl mice following single ip injections. Chlorthiamid (12.25, and 50 mg/kg) induced an extensive destruction of the olfactory
C Eriksson et al.
Toxicology letters, 76(3), 203-208 (1995-04-01)
Chlorthiamid (2,6-dichlorothiobenzamide) and its major metabolite 2,6-dichlorobenzonitrile are olfactory toxicants with a high in vivo covalent binding in the olfactory mucosa of mice. This study showed that the cytochrome P450 (P450) inhibitors, metyrapone and sodium-diethyldithiocarbamate, abolished the chlorthiamid-induced toxicity (12

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service