Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

88792

Supelco

2,4,5-Trichloro-6-hydroxyisophthalonitrile

PESTANAL®, analytical standard

Synonym(s):

2,4,5-Trichloro-6-hydroxy-1,3-benzenedicarbonitrile, Hydroxychlorothalonil

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8HCl3N2O
CAS Number:
Molecular Weight:
247.47
Beilstein:
2653044
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

PESTANAL®

Assay

≥98.0% (HPLC)

shelf life

limited shelf life, expiry date on the label

application(s)

agriculture
environmental

format

neat

storage temp.

2-8°C

SMILES string

ClC1=C(C#N)C(Cl)=C(Cl)C(O)=C1C#N

InChI

1S/C8HCl3N2O/c9-5-3(1-12)6(10)7(11)8(14)4(5)2-13/h14H

InChI key

MDQKYGOECVSPIW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 2 Inhalation - Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 2 - Eye Dam. 1 - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Robert Green et al.
FEMS microbiology letters, 365(19) (2018-09-06)
Cytochrome P450s have been shown to play a vital role in the xenobiotic detoxification system of Sclerotinia homoeocarpa, the causal agent of the turfgrass disease dollar spot. A previous study indicated that three CYP450s were validated to play a functional

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service