Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

44987

Supelco

Ginsenoside Rc

analytical standard

Synonym(s):

(3β,12β)-20-[(6-O-α-L-Arabinofuranosyl-β-D-glucopyranosyl)oxy]-12-hydroxydammar-24-en-3-yl 2-O-β-D-glucopyranosyl-β-D-glucopyranoside, NSC 310104, Panaxoside Rc

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C53H90O22
CAS Number:
Molecular Weight:
1079.27
EC Number:
MDL number:
UNSPSC Code:
85151701
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥98.0% (HPLC)

shelf life

limited shelf life, expiry date on the label

application(s)

food and beverages

format

neat

storage temp.

2-8°C

SMILES string

C[C@@]12CC[C@@]3([H])C(C)(C)[C@@H](O[C@]4([H])O[C@H](CO)[C@@H](O)[C@H](O)[C@H]4O[C@@]5([H])[C@H](O)[C@@H](O)[C@H](O)[C@@H](CO)O5)CC[C@]3(C)[C@@]1([H])C[C@@H](O)[C@@]6([H])[C@]2(CC[C@]6([H])[C@](CCC=C(C)C)(O[C@@H]7O[C@H](CO[C@@H]8O[C@@H](CO)[C@H](O)[C@H]8O

InChI

1S/C53H90O22/c1-23(2)10-9-14-53(8,75-47-43(67)39(63)37(61)29(72-47)22-68-45-41(65)36(60)28(21-56)69-45)24-11-16-52(7)33(24)25(57)18-31-50(5)15-13-32(49(3,4)30(50)12-17-51(31,52)6)73-48-44(40(64)35(59)27(20-55)71-48)74-46-42(66)38(62)34(58)26(19-54)70-46/h10,24-48,54-67H,9,11-22H2,1-8H3

InChI key

JDCPEKQWFDWQLI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Ginsenoside Rc, a bioactive constituent of plant genus Panax ginseng, belongs to the class of protopanaxadiol-type saponins. It can play an important role in disease prevention owing to its potential physiological activity.

Application

Ginsenoside Rc may be used as an analytical standard for the determination of the analyte in herbal products/extracts containing Panax ginseng (Korean or Chinese ginseng) and P. quinquefolius (American ginseng), biological samples, and milk beverages by liquid chromatography (LC) based techniques.

Other Notes

This compound is commonly found in plants of the genus: panax

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Electrospray Liquid Chromatography/Mass Spectrometry of Ginsenosides
van Breeman, et al.
Analytical Chemistry, 67, 3985-3989 (1995)
A Tárrega et al.
Journal of dairy science, 95(8), 4246-4255 (2012-07-24)
American ginseng (Panax quinquefolius L.) has recognized neurocognitive effects, and a ginsenoside-rich extract of the root of the plant has been shown to improve cognitive functions in young adults. This study aimed at assessing the chemical and sensory profiles of
Myoung-Su Lee et al.
Journal of ethnopharmacology, 127(3), 771-776 (2009-12-08)
Panax ginseng and its major component, ginsenosides, are widely used for the prevention of various disorders in oriental medicine. To evaluate the effect of ginsenoside Rc (Rc), one of the active constituents in Panax ginseng, on glucose uptake in C2C12
Q C Ji et al.
Phytochemical analysis : PCA, 12(5), 320-326 (2001-11-14)
An HPLC-MS/MS method was developed for the quantitative determination of ginsenosides, which are the marker compounds for herbal products containing Panax ginseng (Korean or Chinese ginseng) and P. quinquefolius (American ginseng). Samples were extracted with BondElut C18 HF extraction columns
Anbao Wang et al.
Phytochemical analysis : PCA, 16(4), 272-277 (2005-07-27)
In order to determine the active ingredients in root extracts of Panax quinquefolius (American ginseng), a gradient HPLC method involving UV photodiode array detection was applied to separate and quantify simultaneously the ginsenosides Rb1, Rb2, Rc, Rd, Re, Rf and

Articles

Optimize HPLC method for ginsenoside separation using a mixture, applying it to American Ginseng root, with conditions and chromatograms shown.

Optimize HPLC method for ginsenoside separation using a mixture, applying it to American Ginseng root, with conditions and chromatograms shown.

Optimize HPLC method for ginsenoside separation using a mixture, applying it to American Ginseng root, with conditions and chromatograms shown.

Optimize HPLC method for ginsenoside separation using a mixture, applying it to American Ginseng root, with conditions and chromatograms shown.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service