Skip to Content
MilliporeSigma

G0777

Ginsenoside-Rb1 from Panax ginseng (Korean ginseng) root

triterpenoid saponin, ≥98% (HPLC)

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C54H92O23
CAS Number:
Molecular Weight:
1109.29
NACRES:
NA.25
PubChem Substance ID:
UNSPSC Code:
12352201
EC Number:
255-532-8
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

plant root (Panax ginseng)

Quality Segment

assay

≥98% (HPLC)

form

powder

technique(s)

HPLC: suitable

impurities

≤10% water (Karl Fischer)

color

white to yellow cast

mp

186 - 188  °C ((367 - 370 °F ))

solubility

methanol: soluble 9.80-10.20 mg/mL, clear, colorless to faintly yellow

storage temp.

2-8°C

SMILES string

C\C(C)=C\CCC(C)(OC1OC(COC2OC(CO)C(O)C(O)C2O)C(O)C(O)C1O)C3CCC4(C)C3C(O)CC5C6(C)CCC(OC7OC(CO)C(O)C(O)C7OC8OC(CO)C(O)C(O)C8O)C(C)(C)C6CCC45C

InChI

1S/C54H92O23/c1-23(2)10-9-14-54(8,77-48-44(69)40(65)37(62)29(74-48)22-70-46-42(67)38(63)34(59)26(19-55)71-46)24-11-16-53(7)33(24)25(58)18-31-51(5)15-13-32(50(3,4)30(51)12-17-52(31,53)6)75-49-45(41(66)36(61)28(21-57)73-49)76-47-43(68)39(64)35(60)27(20-56)72-47/h10,24-49,55-69H,9,11-22H2,1-8H3/t24-,25+,26+,27+,28+,29+,30-,31+,32-,33-,34+,35+,36+,37+,38-,39-,40-,41-,42+,43+,44+,45+,46+,47-,48-,49-,51-,52+,53+,54-/m0/s1

InChI key

GZYPWOGIYAIIPV-JBDTYSNRSA-N

Application

Ginsenoside-Rb1 from Panax ginseng, a triterpenoid saponin, may be used in ginseng-derived bioactive molecules research to study its mechanisms of action, pharmacokinetics, safety and efficacy as a cardioprotective agent. Ginsenoside-Rb1 may be used to help identify and characterize enzymes with ginsenoside converting activities.

Biochem/physiol Actions

Triterpene saponin found in ginseng.

Other Notes

To gain a comprehensive understanding of our extensive range of Polysaccharides for your research, we encourage you to visit our Carbohydrates Category page.


Pictograms

Exclamation mark

Signal Word

Warning

Hazard Codes

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class

11 - Combustible Solids

WGK

WGK 3

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable

PPE (personal protective equipment)

dust mask type N95 (US), Eyeshields, Gloves



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library