Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

44948

Supelco

Iron(II) gluconate hydrate

Synonym(s):

Ferrous gluconate hydrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C6H11O7)2Fe · aq
CAS Number:
Molecular Weight:
446.14 (anhydrous basis)
Beilstein:
3805721
MDL number:
UNSPSC Code:
12000000
PubChem Substance ID:
NACRES:
NA.21

Quality Level

loss

5-15% loss on drying

anion traces

chloride (Cl-): ≤500 mg/kg
sulfate (SO42-): ≤500 mg/kg

SMILES string

[H]O[H].OC[C@@H](O)[C@@H](O)[C@H](O)[C@@H](O)C(=O)O[Fe]OC(=O)[C@H](O)[C@@H](O)[C@H](O)[C@H](O)CO

InChI

1S/2C6H12O7.Fe.H2O/c2*7-1-2(8)3(9)4(10)5(11)6(12)13;;/h2*2-5,7-11H,1H2,(H,12,13);;1H2/q;;+2;/p-2/t2*2-,3-,4+,5-;;/m11../s1

InChI key

BJBSKTPEFSQVHL-XRDLMGPZSA-L

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Bhawana Sachdeva et al.
International journal for vitamin and nutrition research. Internationale Zeitschrift fur Vitamin- und Ernahrungsforschung. Journal international de vitaminologie et de nutrition, 91(1-2), 133-142 (2019-11-09)
The objective of this investigation was to evaluate the stability of vitamin A in fortified milk which was exposed to different processing conditions viz. heat treatments (pasteurization, boiling and sterilization), light exposure treatments (1485, 2970 and 4455 lux for 2, 4

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service