Skip to Content
MilliporeSigma
All Photos(2)

Documents

44253

Supelco

Dotriacontane

analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)30CH3
CAS Number:
Molecular Weight:
450.87
Beilstein:
1783260
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

vapor density

15.6 (vs air)

vapor pressure

<1 mmHg ( 20 °C)

Assay

≥98.0% (GC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

bp

467 °C (lit.)

mp

65-70 °C (lit.)
69-71 °C

application(s)

petroleum

format

neat

SMILES string

CCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCC

InChI

1S/C32H66/c1-3-5-7-9-11-13-15-17-19-21-23-25-27-29-31-32-30-28-26-24-22-20-18-16-14-12-10-8-6-4-2/h3-32H2,1-2H3

InChI key

QHMGJGNTMQDRQA-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Dotriacontane is classified under the non-polar organic group of compounds.

Application

Dotriacontane may be used as an analytical reference standard for the determination of the analyte in aerosol samples by various chromatographic techniques.
Dotriacontane may be used as an analytical standard for the determination of Fusarium mycotoxin, fusaproliferin in corn using gas chromatography coupled with flame ionization detection.
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A Gas Chromatography- Flame Ionization Detection Method for Detection of Fusaproliferin in Corn.
Wu X and Smith JS
Journal of Agricultural and Food Chemistry, 55(8), 3211- 3216 (2007)
In-injection port thermal desorption and subsequent gas chromatography-mass spectrometric analysis of polycyclic aromatic hydrocarbons and n-alkanes in atmospheric aerosol samples
Ho SSH and Yu JZ
Journal of Chromatography A, 1059(1-2), 121-129 (2004)
Precautions for in-injection port thermal desorption-gas chromatography/mass spectrometry (TD-GC/MS) as applied to aerosol filter samples.
Ho SSH, et al.
Atmospheric Environment, 45(7), 1491-1496 (2011)
Evaluation of an in-injection port thermal desorption-gas chromatography/mass spectrometry method for analysis of non-polar organic compounds in ambient aerosol samples.
Ho SSH, et al.
Journal of Chromatography A, 1200(2), 217-227 (2008)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service