Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

87087

Sigma-Aldrich

Tetracontane

≥95.0% (GC)

Synonym(s):

n-Tetracontane

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)38CH3
CAS Number:
Molecular Weight:
563.08
Beilstein:
1800152
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

≥95.0% (GC)

form

flakes

mp

80-84 °C (lit.)

SMILES string

CCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCC

InChI

1S/C40H82/c1-3-5-7-9-11-13-15-17-19-21-23-25-27-29-31-33-35-37-39-40-38-36-34-32-30-28-26-24-22-20-18-16-14-12-10-8-6-4-2/h3-40H2,1-2H3

InChI key

KUPLEGDPSCCPJI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Tetracontane is a long chain n-alkane that has been used as a prototype of linear polyethylene to understand the phase behavior of polymer solution.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Melting and crystallization of ultra-high-molecular-weight polyethylene in a mixture with tetracontane under high pressure.
Nakafuku C, et al.
Polymer, 32(4), 696-702 (1991)
Temperature-and pressure-induced crystallization and melting of tetracontane in propane: evidence of retrograde crystallization.
Luszczyk M & Radosz, M
Journal of Chemical and Engineering Data, 48(2), 226-230 (2033)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service