Skip to Content
MilliporeSigma

43883

Octamethylcyclotetrasiloxane

analytical standard

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
[-Si(CH3)2O-]4
CAS Number:
Molecular Weight:
296.62
UNSPSC Code:
77101502
NACRES:
NA.24
PubChem Substance ID:
EC Number:
209-136-7
Beilstein/REAXYS Number:
1787074
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

analytical standard

Quality Segment

vapor density

1 (vs air)

assay

≥97.0% (GC)

form

liquid

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable, gas chromatography (GC): suitable

refractive index

n20/D 1.396 (lit.)

bp

175-176 °C (lit.)

mp

17-18 °C (lit.)

density

0.956 g/mL at 25 °C (lit.)

application(s)

cleaning products
cosmetics
environmental
food and beverages
personal care

format

neat

SMILES string

C[Si]1(C)O[Si](C)(C)O[Si](C)(C)O[Si](C)(C)O1

InChI

1S/C8H24O4Si4/c1-13(2)9-14(3,4)11-16(7,8)12-15(5,6)10-13/h1-8H3

InChI key

HMMGMWAXVFQUOA-UHFFFAOYSA-N

General description

This compound is listed in the SVHC (Substances of very high concern) candidate list of ECHA (European Chemicals Agency)

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.


Still not finding the right product?

Explore all of our products under Octamethylcyclotetrasiloxane


Storage Class

3 - Flammable liquids

flash_point_f

123.8 °F - closed cup

flash_point_c

51 °C - closed cup

signalword

Danger

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Flam. Liq. 3 - PBT - Repr. 2 - vPvB



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Nicholas A Warner et al.
Environmental pollution (Barking, Essex : 1987), 190, 109-114 (2014-04-22)
Spatial distribution and relationship of allometric measurements (length, weight and age) to liver concentrations of cyclic volatile methyl siloxanes (cVMS) including octamethylcyclotetrasiloxane (D4), decamethylcyclopentasiloxane (D5) and dodecamethylcyclosiloxane (D6) in Atlantic cod (Gadus morhua) collected near the community of Tromsø in



Global Trade Item Number

SKUGTIN
43883-100MG04061832245850