MilliporeSigma
  • Allometric relationships to liver tissue concentrations of cyclic volatile methyl siloxanes in Atlantic cod.
Allometric relationships to liver tissue concentrations of cyclic volatile methyl siloxanes in Atlantic cod.
Environmental pollution (Barking, Essex : 1987) (2014-04-22)
Nicholas A Warner, Therese H Nøst, Hector Andrade, Guttorm Christensen
ABSTRACT
Spatial distribution and relationship of allometric measurements (length, weight and age) to liver concentrations of cyclic volatile methyl siloxanes (cVMS) including octamethylcyclotetrasiloxane (D4), decamethylcyclopentasiloxane (D5) and dodecamethylcyclosiloxane (D6) in Atlantic cod (Gadus morhua) collected near the community of Tromsø in Northern Norway were assessed. These congeners were benchmarked against known persistent polychlorinated biphenyls (PCBs 153 and 180) to assess accumulation behavior of cVMS. D5 was the dominate cVMS detected in all fish livers with lipid normalized concentrations up to 10 times or greater than those observed for PCB 153 and 180. D4 and D6 concentration were negatively correlated with fish length and weight, indicating a greater elimination capacity compared to uptake processes with increasing fish size for these chemicals. These results indicate relationships between allometric measurements and cVMS concentrations may account for concentration variations observed within fish and should be assessed in future studies evaluating cVMS bioaccumulation potential.
Product Description
Supelco
Octamethylcyclotetrasiloxane, analytical standard
Sigma-Aldrich
Octamethylcyclotetrasiloxane, 98%
Supelco
PCB No 153, analytical standard
2,2′,4,4′,5,5′-Hexachlorobiphenyl (IUPAC No. 153), BCR®, certified reference material