Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

431419

Supelco

Phosphorus pentoxide

≥99.99% trace metals basis

Synonym(s):

Phosphoric anhydride, Phosphorus(V) oxide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
P2O5
CAS Number:
Molecular Weight:
141.94
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
NACRES:
NA.21

vapor density

4.9 (vs air)

Quality Level

vapor pressure

1 mmHg ( 384 °C)
10 mmHg ( 238 °C)

Assay

≥99.99% trace metals basis

form

powder

impurities

≤0.02% insolubles
<100 ppm total metallic impurities

mp

340 °C (lit.)

density

2.3 g/mL at 25 °C (lit.)

cation traces

NH4+: ≤0.01%
heavy metals: ≤0.01%

SMILES string

O=P12OP3(=O)OP(=O)(O1)OP(=O)(O2)O3

InChI

1S/O10P4/c1-11-5-12(2)8-13(3,6-11)10-14(4,7-11)9-12

InChI key

YWEUIGNSBFLMFL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Phosphorus pentoxide (P2O5) is a commonly used drying agent. It is a component of Eatons reagent, which is commonly used as a catalyst for dehydration reactions.

Application

Phosphorus pentoxide may be used in the synthesis of the following:
  • Hydroxyapatite (HA) sub-micron fibers.
  • Fluorescent carbon nanoparticles (FCNPs).
  • Bistetramethylammonium (TMA) hydrogencyclotriphosphate and rubidium cyclotetraphosphate using dimethyl sulfoxide as a solvent at low temperature condition.
  • Ethylene glycol methacrylate phosphate (EGMP), a UV-reactive phosphonic acid.

Features and Benefits

Meets A.C.S. reagent specifications.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1A

Supplementary Hazards

Storage Class Code

8B - Non-combustible corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Nayara Koba de Moura et al.
Materials science & engineering. C, Materials for biological applications, 117, 111327-111327 (2020-09-14)
Polymer membranes have been widely used in guided bone regeneration (GBR), especially when it comes to their use in dentistry. Poly (lactic acid) (PLA) have good mechanical properties such as flexibility, which allows the material to be moldable and also
One-step spontaneous synthesis of fluorescent carbon nanoparticles with thermosensitivity from polyethylene glycol.
Li S, et al.
New. J. Chem., 39(9), 7033-7039 (2015)
Flame retardation behaviors of UV-curable phosphorus-containing PU coating system.
Wang SC, et al.
Journal of Polymer Engineering, 33(8), 749-756 (2013)
Development of matrix dispersion transdermal therapeutic system containing glipizide.
Behin SR, et al.
Der Pharmacia Lettre, 5, 278-286 (2013)
Electrospun hydroxyapatite fibers from a simple sol-gel system.
Franco PQ, et al.
Materials Letters, 67(1), 233-236 (2012)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service