Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

42078

Supelco

mono-[(2RS)-2-Ethyl-5-oxohexyl] phthalate

analytical standard

Synonym(s):

1,2-Benzenedicarboxylic acid 1-(2-ethyl-5-oxohexyl) ester, mono-(2-Ethyl-5-oxohexyl) phthalate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H20O5
CAS Number:
Molecular Weight:
292.33
Beilstein:
920863
MDL number:
UNSPSC Code:
77101502
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥95.0% (HPLC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

refractive index

n20/D 1.516-1.520

application(s)

cleaning products
cosmetics
environmental
food and beverages
personal care

format

neat

storage temp.

2-8°C

SMILES string

O=C(OCC(CC)CCC(C)=O)C1=CC=CC=C1C(O)=O

InChI

1S/C16H20O5/c1-3-12(9-8-11(2)17)10-21-16(20)14-7-5-4-6-13(14)15(18)19/h4-7,12H,3,8-10H2,1-2H3,(H,18,19)

InChI key

HCWNFKHKKHNSSL-UHFFFAOYSA-N

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yingshuang Zhu et al.
Environment international, 88, 67-73 (2016-01-02)
Total urinary phthalate metabolites (the free plus glucuronidated forms) have been frequently measured in the general population. However, data are limited on the free forms which may be more bioactive, especially for sensitive population such as pregnant women. Here the
Yeni Kim et al.
Environmental health perspectives, 119(10), 1495-1500 (2011-07-09)
There are increasing concerns over adverse effects of prenatal phthalate exposure on the neurodevelopment of infants. Our goal was to explore the association between prenatal di(2-ethylhexyl) phthalate and dibutyl phthalate exposure and the Mental and Psychomotor Developmental Indices (MDI and
Johanna Inhyang Kim et al.
Journal of attention disorders, 25(2), 161-170 (2018-05-22)
Objective: We investigated the interaction effect between the methylation of dopamine receptor D4 (DRD4) and phthalate exposure in ADHD on continuous performance test (CPT) variables. Method: Urine concentrations of mono-(2-ethyl-5-hydroxyhexyl) phthalate (MEHHP), mono-(2-ethyl-5-oxohexyl) phthalate (MEOHP), and mono-n-butyl phthalate (MBP) were
Jacob Bejder et al.
Medicine and science in sports and exercise, 53(1), 236-243 (2020-07-23)
Autologous blood transfusion is performance enhancing and prohibited in sport but remains difficult to detect. This study explored the hypothesis that an untargeted urine metabolomics analysis can reveal one or more novel metabolites with high sensitivity and specificity for detection
Soo-Churl Cho et al.
Environmental health perspectives, 118(7), 1027-1032 (2010-03-03)
Concern over phthalates has emerged because of their potential toxicity to humans. We investigated the relationship between the urinary concentrations of phthalate metabolites and children's intellectual functioning. This study enrolled 667 children at nine elementary schools in five South Korean

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service