Skip to Content
MilliporeSigma
All Photos(1)

Documents

41159

Millipore

Actinomyces Agar, Vegitone

suitable for microbiology, NutriSelect® Plus

Synonym(s):

Vegitone Actinomyces Agar

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41171605
NACRES:
NA.85

sterility

non-sterile

Quality Level

form

powder

shelf life

limited shelf life, expiry date on the label

composition

agar, 20 g/L
ammonium sulfate, 1 g/L
calcium chloride, 0.02 g/L
L-cysteine hydrochloride, 1 g/L
dextrose, 5 g/L
magnesium sulfate, 0.2 g/L
monopotassium phosphate, 15 g/L
sodium chloride, 5 g/L
starch, soluble, 1 g/L
tryptone (vegetable), 4 g/L
tryptose (vegetable), 10 g/L
vegetable infusions powder, 10 g/L
yeast extract, 5 g/L

manufacturer/tradename

NutriSelect® Plus

technique(s)

microbiological culture: suitable

final pH

6.9±0.2 (25 °C)

application(s)

agriculture
clinical testing
environmental
food and beverages
life science and biopharma
veterinary

microbiology

suitability

Actinomycetes spp.
Streptomyces spp.

General description

Actinomyces media are recommended for the cultivation and maintenance of the anaerobic Actinomyces species. This medium contains no animal derived material.

Preparation Note

Suspend 77.22 g in 1000 ml distilled water. Heat to boiling to dissolve the medium completely. Distribute into tubes or flasks. Sterilize by autoclaving at 15 lbs pressure (121°C) for 15 minutes.

Footnote

We offer two media types: the superior granulated GranuCult® and the cost-efficient powdered NutriSelect® culture media, depending on your needs.
The designations basic, plus, or prime are added to indicate the quality control level, from basic quality control to standard QC plus to prime for full regulatory compliance.

Legal Information

GRANUCULT is a registered trademark of Merck KGaA, Darmstadt, Germany
NutriSelect is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service