Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

394904

Supelco

Pentafluoropropionic anhydride

for GC derivatization, LiChropur, 99%

Synonym(s):

PFPA, Perfluoropropionic anhydride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CF3CF2CO)2O
CAS Number:
Molecular Weight:
310.05
Beilstein:
1806446
EC Number:
MDL number:
UNSPSC Code:
12000000
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

grade

for GC derivatization

Quality Level

Assay

99%

form

liquid

quality

LiChropur

reaction suitability

reagent type: derivatization reagent
reaction type: Acylations

technique(s)

gas chromatography (GC): suitable

bp

69-70 °C/735 mmHg (lit.)

density

1.571 g/mL at 25 °C (lit.)

SMILES string

FC(F)(F)C(F)(F)C(=O)OC(=O)C(F)(F)C(F)(F)F

InChI

1S/C6F10O3/c7-3(8,5(11,12)13)1(17)19-2(18)4(9,10)6(14,15)16

InChI key

XETRHNFRKCNWAJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Learn more in the Product Information
Derivatization reagent used in a GC-MS analysis of stereoisomer content of synthetic peptides.[1] Also used in a GC-MS detection method for synephrine in dietary supplements.[2]
For the preparation of volatile pentafluoropropionyl derivatives of a wide range of compounds, for example, amines and alcohols, for EC/GC.
Pentafluoropropionic anhydride (PFPA) was used as derivatization reagent for urine samples to determine the presence of codeine, morphine, 6-monoacetylmorphine, ethylmorphine, and dihydrocodeine using GC-MS in selected ion monitoring (SIM) mode.[3] It was also used to derivatize the dry pericardial fluid sample extracts for quantification of cocaine, benzoylecgonine, and cocaethylene in pericardial fluid using GC-MS.[4]

Packaging

Clear borosilicate glass, high hydrolytic resistance (Type I)

Legal Information

LiChropur is a trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Skin Corr. 1B

Supplementary Hazards

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ralf Pätzold et al.
Chirality, 18(7), 551-557 (2006-05-10)
Synthetic dipeptides comprising mixtures of enantiomers, diastereomers, or sequential isomers were converted into their N-perfluoroacetyl dipeptide esters (perfluoroacetyl: trifluoroacetyl, pentafluoroacetyl, heptafluorobutyryl; ester: methyl, 1-propyl, 2-propyl, 2,2,2-trifluoroethyl) and analyzed by GC-MS on the chiral stationary phases Chirasil-L-Val and Lipodex-E using helium
Emilia Marchei et al.
Journal of pharmaceutical and biomedical analysis, 41(4), 1468-1472 (2006-05-16)
A simple and rapid procedure based on gas chromatography-mass spectrometry (GC-MS) is described for determination of synephrine, active principle of Citrus aurantium plant, in solid and liquid dietary supplements. After the addition of 3,4-methylenedioxypropylamphetamine as internal standard (I.S.), a liquid-liquid
Comparison of RapiTest? with Emit? d, au? and GC-MS for the Analysis of Drugs in Urine.
Korte, T., et al.
Journal of Analytical Toxicology, 21, 49-53 (1997)
F F Hsu et al.
Analytical biochemistry, 216(2), 401-405 (1994-02-01)
We report an isotope dilution mass spectrometry method using capillary gas chromatography negative ion chemical ionization for the determination of cortisol in human plasma. Cortisol was derivatized with pentafluoropropionic anhydride at room temperature and measured by selected ion monitoring of
Y S Oh-Shin et al.
Journal of chromatography. A, 769(2), 285-291 (1997-05-09)
A rapid and simple gas chromatographic method for the simultaneous determination of malathion, parathion, fenitrothion, diazinon and carbaryl in drinking water is presented. A fused-silica SE-54 column was used for the separation of the insecticides and was split into two

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service