Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

556688

Sigma-Aldrich

Dimethoxydimethylsilane

≥99.5%, 99.999% metals basis

Synonym(s):

Dimethyldimethoxysilane

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Si(OCH3)2(CH3)2
CAS Number:
Molecular Weight:
120.22
Beilstein:
1733694
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

Assay

≥99.5%
99.999% metals basis

form

liquid

refractive index

n20/D 1.369 (lit.)

bp

81.4 °C (lit.)

density

0.88 g/mL at 25 °C (lit.)

SMILES string

CO[Si](C)(C)OC

InChI

1S/C4H12O2Si/c1-5-7(3,4)6-2/h1-4H3

InChI key

JJQZDUKDJDQPMQ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Flame

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

50.0 °F - closed cup

Flash Point(C)

10 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

C Wyatt Shields et al.
Journal of controlled release : official journal of the Controlled Release Society, 278, 37-48 (2018-04-01)
Retinol, a derivative of vitamin A, is a ubiquitous compound used to treat acne, reduce wrinkles and protect against conditions like psoriasis and ichthyosis. While retinol is used as the primary active ingredient (AI) in many skin care formulations, its
Chiara Micalella et al.
Biotechnology and applied biochemistry, 61(3), 297-303 (2014-02-28)
Enzymes entrapped in wet, nanoporous silica gel have great potential as bioreactors for bioremediation because of their improved thermal, chemical, and mechanical stability with respect to enzymes in solution. The B isozyme of catechol 1,2 dioxygenase from Acinetobacter radioresistens and

Articles

atomic layer deposition (ALD), microelectronics, Mo:Al2O3 films, nanocomposite coating, photovoltaics, semiconductor devices, W:Al2O3 films, composite films, layer-by-layer

Hybrid organic-inorganic sol-gel materials containing silica were first called “ORMOSILs” in 1984.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service