Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

34344

Supelco

Flutriafol

PESTANAL®, analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H13F2N3O
CAS Number:
Molecular Weight:
301.29
Beilstein:
8155287
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

SMILES string

OC(Cn1cncn1)(c2ccc(F)cc2)c3ccccc3F

InChI

1S/C16H13F2N3O/c17-13-7-5-12(6-8-13)16(22,9-21-11-19-10-20-21)14-3-1-2-4-15(14)18/h1-8,10-11,22H,9H2

InChI key

JWUCHKBSVLQQCO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Flutriafol is a sterol-inhibiting triazole fungicide. Its mode of action is through the inhibition of C-14α-demethylase enzyme involved in fungal sterol biosynthesis.

Application

Flutriafol may be used as a reference standard in the determination of flutriafol in food and environmental matrices by reversed-phase high-performance liquid chromatography.
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Simultaneous enantioselective determination of triazole fungicide flutriafol in vegetables, fruits, wheat, soil, and water by reversed-phase high-performance liquid chromatography.
Zhang Q, et al.
Journal of Agricultural and Food Chemistry, 62(13), 2809-2815 (2014)
Biological mode of action of soil-applied flutriafol in controlling take-all of wheat.
Cotterill PJ.
Soil Biology and Biochemistry, 23(4), 323-329 (1991)
Yuba R Kandel et al.
Plant disease, 104(6), 1736-1743 (2020-04-15)
In total, 52 uniform field experiments were conducted in Illinois, Indiana, Iowa, Michigan, South Dakota, and Wisconsin in the United States and Ontario, Canada from 2013 to 2017 comparing crop protection products against sudden death syndrome (SDS) of soybean. Data
H L Mehl et al.
Plant disease, 104(2), 438-447 (2019-12-11)
Fungicide programs for managing target spot of cotton caused by Corynespora cassiicola were evaluated over 15 site-years in the southeastern United States between 2014 and 2016. Two cultivars, hypothesized to vary in target spot susceptibility, PhytoGen 499WRF (PHY499) and Deltapine
Heather Davies-Strickleton et al.
PloS one, 15(2), e0229414-e0229414 (2020-02-29)
Onychomycosis, or fungal nail infection, is a common fungal infection largely caused by dermatophyte fungi, such as Trichophyton rubrum or Trichophyton mentagrophytes, which affects a significant number of people. Treatment is either through oral antifungal medicines, which are efficacious but

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service