Skip to Content
MilliporeSigma
All Photos(1)

Documents

36428

Supelco

Keracyanin chloride

analytical standard

Synonym(s):

Cyanidin-3-O-rutinoside chloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C27H31ClO15
CAS Number:
Molecular Weight:
630.98
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥98% (HPLC)

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

food and beverages

format

neat

storage temp.

−20°C

SMILES string

[Cl-].C[C@@H]1O[C@@H](OC[C@H]2O[C@@H](Oc3cc4c(O)cc(O)cc4[o+]c3-c5ccc(O)c(O)c5)[C@H](O)[C@@H](O)[C@@H]2O)[C@H](O)[C@H](O)[C@H]1O

InChI

1S/C27H30O15.ClH/c1-9-19(32)21(34)23(36)26(39-9)38-8-18-20(33)22(35)24(37)27(42-18)41-17-7-12-14(30)5-11(28)6-16(12)40-25(17)10-2-3-13(29)15(31)4-10;/h2-7,9,18-24,26-27,32-37H,8H2,1H3,(H3-,28,29,30,31);1H/t9-,18+,19-,20+,21+,22-,23+,24+,26+,27+;/m0./s1

InChI key

ADZHXBNWNZIHIX-XYGAWYNKSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Keracyanin chloride is an anthocyanin standard, which may be used for the liquid chromatography-mass spectrometry (LC-MS) determination and anti-microbial analysis of grape pomace, the solid residue of grapes in the winemaking process.
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M Miskulin et al.
Journal francais d'ophtalmologie, 7(11), 737-743 (1984-01-01)
Cyaninoside chloride is a drug used in the treatment of visual functional troubles in low-luminance conditions, in patients suffering from myopia and night blindness. Its effects on the enzymatic degradation of collagen by collagenase have been studied by means of
Patiwit Loypimai et al.
Journal of food science and technology, 53(1), 461-470 (2016-01-21)
The study of the stability of anthocyanins in food colorant powder is important to predict the quality changes occurring as the food products are processed, to prevent and control the degradation of the anthocyanins. The objectives of this study were
Narangerel Altangerel et al.
Proceedings of the National Academy of Sciences of the United States of America, 114(13), 3393-3396 (2017-03-16)
Development of a phenotyping platform capable of noninvasive biochemical sensing could offer researchers, breeders, and producers a tool for precise response detection. In particular, the ability to measure plant stress in vivo responses is becoming increasingly important. In this work
R Márquez et al.
Pharmacology & toxicology, 89(5), 255-258 (2002-03-08)
Exercise generates free radicals only when it is exhaustive. Free radicals are involved in tissue damage caused by exercise. Antioxidant vitamins (vitamin C and E) and other antioxidants such as coenzyme Q, and N-acetyl cysteine prevent muscle damage and decrease
P Solé et al.
Journal francais d'ophtalmologie, 7(1), 35-39 (1984-01-01)
A controlled, clinical trial, comparing cyaninoside chloride and Heleniene , was conducted on 31 out-patients suffering from functional disturbances of vision in low-luminance conditions. The evolution of photopic and mesopic visual acuities, electro- oculograms and adapto -electroretinograms was assessed for

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service