Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

19050

Sigma-Aldrich

Idaein chloride

≥90% (HPLC)

Synonym(s):

3-(Galactosyloxy)-3′,4′,5,7-tetrahydroxyflavylium chloride, Cyanidin 3-O-β-D-galactopyranoside, Cyanidin 3-O-galactoside chloride, Idein chloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H21ClO11
CAS Number:
Molecular Weight:
484.84
MDL number:
UNSPSC Code:
12352201
PubChem Substance ID:
NACRES:
NA.25

Quality Level

Assay

≥90% (HPLC)

form

powder or crystals

color

red to dark brown

storage temp.

−20°C

SMILES string

[Cl-].OC[C@H]1O[C@@H](Oc2cc3c(O)cc(O)cc3[o+]c2-c4ccc(O)c(O)c4)[C@H](O)[C@@H](O)[C@H]1O

InChI

1S/C21H20O11.ClH/c22-7-16-17(27)18(28)19(29)21(32-16)31-15-6-10-12(25)4-9(23)5-14(10)30-20(15)8-1-2-11(24)13(26)3-8;/h1-6,16-19,21-22,27-29H,7H2,(H3-,23,24,25,26);1H/t16-,17+,18+,19-,21-;/m1./s1

InChI key

YTMNONATNXDQJF-QSLGVYCOSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Idaein chloride (Cyanidin 3-O-galactoside) is a glycoside derivative of cyanidin, a potent inhibitor of the epidermal growth-factor receptor, used in comparative analysis of anti-EGF activity in cancer cells.

Biochem/physiol Actions

Anthocyanin.

Other Notes

To gain a comprehensive understanding of our extensive range of Monosaccharides for your research, we encourage you to visit our Carbohydrates Category page.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

K Briviba et al.
Nutrition and cancer, 41(1-2), 172-179 (2002-07-04)
Dietary polyphenols, including anthocyanidins and their glycosides anthocyanins, are suggested to be involved in the protective effects of fruits and vegetables against cancer. Very few data are available concerning the effects of anthocyanidins/anthocyanins on cellular processes induced by growth factors
Houhua Li et al.
Planta, 226(5), 1243-1254 (2007-07-10)
Flavonoids are a large family of polyphenolic compounds with manifold functions in plants. Present in a wide range of vegetables and fruits, flavonoids form an integral part of the human diet and confer multiple health benefits. Here, we report on
Tarun Belwal et al.
Food chemistry, 297, 124993-124993 (2019-06-30)
Ultrasonic effect on extraction of cyanidin-3-galactoside (Cy3-gal) from pear fruit peel was investigated and compared with conventional extraction (CE) method. Different process factors were tested to determine the optimum conditions for ultrasonic-assisted extraction (UAE). Results revealed that under optimized UAE
Kaisu R Riihinen et al.
Fitoterapia, 97, 78-86 (2014-06-01)
The antimicrobial activity of lingonberry (Vaccinium vitis-idaea L.) was evaluated against two oral pathogens, Streptococcus mutans and Fusobacterium nucleatum. Long-bed gel permeation chromatography (GPC; Sephadex LH-20) yielded purified flavonoids, with the most efficient minimum inhibitory concentrations (MICs) against planktonic cells
Dragan Amić et al.
Bioorganic & medicinal chemistry, 18(1), 28-35 (2009-12-01)
The applicability of the newly developed RM1 and PM6 methods implemented in the semiempirical quantum chemistry mopac2009 software package in modeling free radical scavenging activity of flavonoids was examined. Bond dissociation enthalpy (BDE) of OH groups could be calculated much

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service