Skip to Content
MilliporeSigma
All Photos(1)

Documents

36155

Supelco

Metazachlor

PESTANAL®, analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H16ClN3O
CAS Number:
Molecular Weight:
277.75
Beilstein:
621550
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
NMR: suitable
gas chromatography (GC): suitable

mp

74-78 °C

suitability

passes test for identity (NMR)

application(s)

agriculture
environmental

format

neat

SMILES string

Cc1cccc(C)c1N(Cn2cccn2)C(=O)CCl

InChI

1S/C14H16ClN3O/c1-11-5-3-6-12(2)14(11)18(13(19)9-15)10-17-8-4-7-16-17/h3-8H,9-10H2,1-2H3

InChI key

STEPQTYSZVCJPV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Metazachlor is an α-chloracetamide derivative which is widely used as a herbicide for weed control in the cultivation of crucifers such as rape.

Application

Metazachlor may be used as an analytical standard for the determination of metazachlor in:
  • Natural water samples by high performance liquid chromatography-electrospray-tandem mass spectrometry (HPLC-ESI-MS/MS).
  • Plant foodstuffs by capillary gas chromatography equipped with electron capture detector (ECD) as well as nitrogen phosphorus detector (NPD) followed by MS analysis in electron impact ionization (EI) mode.
  • Drinking and related waters by solid phase extraction (SPE) followed by GC-EI-MS.
  • Wastewater samples by SPE and subsequent analysis with ultra-performance liquid chromatography combined with time of flight mass spectrometry (UPLC-TOF-MS).

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 2 - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Ewa Szpyrka et al.
Scientific reports, 10(1), 3747-3747 (2020-03-01)
The study concerned dissipation of metazachlor and clomazone, herbicides widely used in rapeseed (Brassica napus L. subsp. napus) protection, applied to the clay soil under field and laboratory conditions. Furthermore, the uptake of these pesticide from soil by rapeseed plants
Response of plankton communities in freshwater pond and stream mesocosms to the herbicide metazachlor
Mohr S, et al.
Environmental Pollution (Barking, Essex : 1987), 152(3), 530-542 (2008)
Hanne Vercampt et al.
Environmental toxicology and chemistry, 36(1), 59-70 (2016-06-28)
In accordance with realistic application approaches, a short-term 1-factorial experiment was set up to investigate the phytotoxic impact of pre-emergent application of the chloroacetamide herbicide metazachlor on Brassica napus. In addition to morphological parameters, the underlying processes that ultimately determine
Kamila Lewicka et al.
Materials (Basel, Switzerland), 13(12) (2020-06-25)
The present study aimed to develop and prepare new polymer/herbicide formulations for their potential application in environment-friendly, controlled release systems of agrochemicals. Selected biodegradable polymers, including L-Lactide/Glycolide/PEG/Terpolymer (PLAGA-PEG-PLAGA) as well as oligosaccharide-based polymers and their blend with terpolymer, were used
Christiane Wiemann et al.
Toxicology, 426, 152282-152282 (2019-08-30)
In a 2-year study the herbicide metazachlor (BAS 479H) was shown to significantly increase the incidence of liver tumours in female Wistar rats at a dietary level of 8000 ppm. As metazachlor is not a genotoxic agent, a series of in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service