Skip to Content
MilliporeSigma
All Photos(2)

Documents

33697

Supelco

Dimethenamide-P

PESTANAL®, analytical standard

Synonym(s):

(S)-2-Chloro-N-(2,4-dimethyl-2-thienyl)-N-(2-methoxy-1-methylethyl)acetamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H18ClNO2S
CAS Number:
Molecular Weight:
275.79
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

storage temp.

2-8°C

SMILES string

COC[C@H](C)N(C(=O)CCl)c1c(C)csc1C

InChI

1S/C12H18ClNO2S/c1-8-7-17-10(3)12(8)14(11(15)5-13)9(2)6-16-4/h7,9H,5-6H2,1-4H3/t9-/m0/s1

InChI key

JLYFCTQDENRSOL-VIFPVBQESA-N

Looking for similar products? Visit Product Comparison Guide

General description

Dimethenamide-P is an amide herbicide, which can be used for the pre-emergence control of annual grasses, small-seeded broadleaves, and sedges in ornamentals. Its mode of action involves the inhibition of long chain fatty acids.

Application

Dimethenamide-P may be used as an analytical reference standard for the quntification of the analyte in environmental samples using liquid chromatography coupled to tandem mass spectrometry (LC-MS/MS).
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Skull and crossbonesEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Skin Sens. 1

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Liquid chromatography-tandem mass spectrometric analysis and regulatory issues of polar pesticides in natural and treated waters.
Kuster M, et al.
Journal of Chromatography A, 1216(3), 520-529 (2009)
Sittig's Handbook of Pesticides and Agricultural Chemicals (2014)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service