MilliporeSigma
  • Degradation and mineralization of 3-chlorobiphenyl by a mixed aerobic bacterial culture.
Degradation and mineralization of 3-chlorobiphenyl by a mixed aerobic bacterial culture.
Applied microbiology and biotechnology (1991-11-01)
F Fava, L Marchetti
ABSTRACT
A mixed bacterial culture obtained from polychlorinated-biphenyl-contaminated river sediments proved capable of degrading 3-chlorobiphenyl (3-CB) under aerobic laboratory conditions. Almost total mineralization of 150 mg/l of 3-CB occurred when, after 3 days of incubation, the mineral medium was supplied with benzoic acid as a carbon source. Two strains of Pseudomonas capable of degrading the substrate to 3-chlorobenzoic acid and a strain of Pseudomonas fluorescens capable of co-metabolizing this metabolite were selected from the mixed culture. A nearly stoichiometric amount of chloride, which defines the percentage of total mineralization, was eliminated during mixed culture growth.
Product Description
Supelco
PCB No 2, analytical standard