Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

34209

Supelco

Pirimicarb-d6

PESTANAL®, analytical standard

Synonym(s):

2-Dimethylamino-5,6-dimethyl-4-pyrimidinyl dimethyl-d6-carbamate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11D6H12N4O2
CAS Number:
Molecular Weight:
244.32
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

mass shift

M+6

SMILES string

[2H]C([2H])([2H])N(C(=O)Oc1nc(nc(C)c1C)N(C)C)C([2H])([2H])[2H]

InChI

1S/C11H18N4O2/c1-7-8(2)12-10(14(3)4)13-9(7)17-11(16)15(5)6/h1-6H3/i5D3,6D3

InChI key

YFGYUFNIOHWBOB-SCPKHUGHSA-N

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Skull and crossbonesEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yi-Ching Choi et al.
Journal of mass spectrometry : JMS, 55(2), e4380-e4380 (2019-06-12)
Detection of pesticide residues in food samples is important for safeguarding food quality and safety. Conventional approaches for detection of pesticides in food samples typically involve labour-intensive and time-consuming sample pretreatment and chromatographic separation. In this study, solid phase micro-extraction

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service