Skip to Content
MilliporeSigma

Skip To

32055

Carbaryl

PESTANAL®, analytical standard

Synonym(s):

1-Naphthyl-N-methylcarbamate

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

Linear Formula:
C10H7OCONHCH3
CAS Number:
Molecular Weight:
201.22
UNSPSC Code:
41116107
NACRES:
NA.24
PubChem Substance ID:
EC Number:
200-555-0
Beilstein/REAXYS Number:
1875862
MDL number:
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

analytical standard

Quality Segment

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable, gas chromatography (GC): suitable

mp

142-146 °C (lit.)

application(s)

agriculture
environmental

format

neat

SMILES string

CNC(=O)Oc1cccc2ccccc12

InChI

1S/C12H11NO2/c1-13-12(14)15-11-8-4-6-9-5-2-3-7-10(9)11/h2-8H,1H3,(H,13,14)

InChI key

CVXBEEMKQHEXEN-UHFFFAOYSA-N

General description

Carbaryl is generally used to induce gene expression in human cells.

Application

Carbaryl has been used as reference standard in HPLC quantification analysis for determination of pesticide residue in water samples.
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
559814
description

PESTANAL®, analytical standard

description

97%

format

neat

format

-

technique(s)

gas chromatography (GC): suitable, HPLC: suitable

technique(s)

-

grade

analytical standard

grade

-

product line

PESTANAL®

product line

-

shelf life

limited shelf life, expiry date on the label

shelf life

-

mp

142-146 °C (lit.)

mp

142-146 °C (lit.)


Still not finding the right product?

Explore all of our products under Carbaryl


Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Acute 1 - Carc. 2

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable

PPE (personal protective equipment)

Eyeshields, Faceshields, Gloves, type P3 (EN 143) respirator cartridges



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

  1. Is this chemical suitable for treatment of tree bark in ordre to avoid a fungus.

    1 answer
    1. This product is intended for use as an analytical standard in the identification and quantification of carbaryl in pesticide residues. It was not formulated for direct application to living systems.

      Helpful?

Reviews

No rating value

Active Filters